Bio-Simplex Media Bottle Assembly Systems, 1000mL

Representative image only.

Bio-Simplex Media Bottle Assembly Systems, 1000mL

Bio-Simplex | Mfr: SA38MM-1L-2F-DB| Item: 5014188

Aseptic, single-use solutions for secure fluid handling

$278.47CAD / Each

$278.47CAD / Each
Product Description

Includes

Two port EZ Top® container closure, 18" C-Flex® tubing (1/8" ID x 1/4" OD) with a plug end and dip tube, 0.2 µm vent filter, and one media bottle.

Key Features

  • Closed-system design minimizes contamination risks
  • Low gas permeability ensures sample integrity
  • Clarity for easy content viewing
  • Volume-graduated design for precise measurements
  • Compatible with sterile tubing welding and sealing

More About this Item

Designed for the bioprocess industry, Bio-Simplex® Media Bottle Assembly Systems provide a reliable, closed-system solution for aseptic sample handling, fluid transfer, and storage. Featuring a two-port EZ Top® container closure made from C-Flex® biopharmaceutical thermoplastic elastomer (TPE), these bottles ensure a seamless and sterile fluid path. The 0.2 µm vent filter supports sterile gas exchange, while the thermal bond between stopper and tubing eliminates contamination risks. Available in sizes ranging from 60 mL to 2 L, these assemblies meet a Sterility Assurance Level (SAL) of 10?6, making them ideal for applications such as media storage, buffer preparation, and aseptic sampling.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Specifications
  • Capacity (mL)1000
  • MaterialPolyethylene Terephthalate Glycol
  • Cap MaterialHDPE
  • Bottle ColorClear
  • DescriptionMedia Bottle Assembly System, 2000 mL, 5/Pk
Reviews
Q&A