Atago MASTER-S/MILLM. Salt/specific gravity refractometer
Representative image only.
Atago MASTER-S/MILLM. Salt/specific gravity refractometer
Atago | Mfr: 2493 MASTER-S/MILLM.| Item: 0294045
Units provide high precision, accurate results
$1,015.68CAD / Each
for an additional
$200.01CAD / Each
for an additional
$321.94CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$1,015.68CAD / Each
Buy It Together
Current Item
Atago MASTER-S/MILLM. Salt/specific gravity refractometer
Atago MASTER-S/MILLM. Salt/specific gravity refractometer
0294045
$1,015.68CAD / Each
+
InnoCal NIST-Traceable Calibration; Refractometer
1710500
$200.01CAD / Each
=
Total Price 1215.69 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Measure salt concentration of brine in salted foods, or salt concentration in aquariums or commercial fisheries
  • 6 month warranty
More About this Item
Salt/Specific Gravity Refractometer measures salt in parts per thousandand specific gravity (1.000 to 1.070) of seawater. Made of polybutylene terepthalate (PBT) thermoplastic.
6 Month WarrantyISO9001 Registered Supplier
Specifications
  • Min Salinity Range (%)0
  • Max Salinity Range (%)100
  • Resolution1 (ppt); 0.001 (s.g.)
  • Accuracy±1 (ppt); ±0.001 (s.g.)
  • DescriptionSalt/specific gravity refractometer