Ashcroft Pressure Switch 0 to 1000 PSI; 1/4" NPT(F)

Representative image only.

Ashcroft Pressure Switch 0 to 1000 PSI; 1/4" NPT(F)

Ashcroft | Mfr: FPSN7PV07 1000PSI| Item: 6835080

Approved for Class I and II hazardous indoor locations

$879.61CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$879.61CAD / Each

Buy It Together

Current Item

Ashcroft Pressure Switch 0 to 1000 PSI; 1/4" NPT(F)
Ashcroft Pressure Switch 0 to 1000 PSI; 1/4" NPT(F)
6835080

$879.61CAD / Each
+ =
Total Price 1144.45 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Enclosure is NEMA 7/9 and IP66 rated
  • Watertight anodized aluminum body for environmental protection
  • Diaphragm-sealed piston actuator offers long, reliable service
  • Hermetically sealed snap-acting switch element
  • Field adjustable
  • Factory sealed leads
  • Setpoint adjustable from 15-100% of range
  • Simple to install and use
  • Class I, Groups A,B,C, or D and Class II, Group E,F, or G
  • 1 year warranty

More About this Item

These Ashcroft F-Series pressure switches are designed to satisfy most requirements for alarm, shutdown, control and interlock applications on a wide variety of process and equipment. Electromechanical pressure switches are usually more cost effective than transmitters for these types of applications. Materials of construction have been selected for long life.

Applications
Hazardous locations
Engines and compressors
Corrosive environments
Process applications
Offshore applications
Pipelines
Machine tools
Replacement and retrofit
Where size is a consideration or equipment is being downsized

Conformite Europeenne1 Year WarrantyCanadian Standards AssociationIntrinsically SafeIP 66 RatedUL Listed.
Specifications
  • Accuracy±1% full-scale
  • Min Pressure (PSI)0
  • Max Pressure (PSI)1000
  • Min Temperature (° F)20
  • Max Temperature (° F)200
  • Min Temperature (° C)-7
  • Max Temperature (DegreeC)93
  • Wetted Materials316 SS, PTFE, Viton®
  • CaseAluminum
  • Process Connection1/2" NPT(F)
  • Battery Type125/250 VAC, 3-wire
  • Electrical Connections1/2" conduit
  • Switch output5 A at 125/250 VAC
  • OutputSPDT
  • Enclosure RatingNEMA 7/9, IP66
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionPressure Switch 0 to 1000 PSI; 1/4" NPT(F)
  • Warranty1 year
Reviews
Q&A