Advantec 07030100 Test Strips, Cyanogen Check (Cyanide Cn-), 0 to 1000 ppm; 100 Strips/Pk

Representative image only.

Advantec 07030100 Test Strips, Cyanogen Check (Cyanide Cn-), 0 to 1000 ppm; 100 Strips/Pk

Advantec | Mfr: 07030100| Item: 5920124

Color-change test papers give rapid indication of pH level ion presence

$107.56CAD / Pkg of 100

$107.56CAD / Pkg of 100

Buy It Together

Current Item

Advantec 07030100 Test Strips, Cyanogen Check (Cyanide Cn-), 0 to 1000 ppm; 100 Strips/Pk
Advantec 07030100 Test Strips, Cyanogen Check (Cyanide Cn-), 0 to 1000 ppm; 100 Strips/Pk
5920124

$107.56CAD / Pkg of 100
+
Cole-Parmer Phosphoric Acid, 85%, ACS Grade, 1 L
8840180

$151.76CAD / Each
+ +
Reagents Conductivity Standard 1413 µS/cm, 1L
7891521

$89.73CAD / Each
+ +
LabChem Nitric Acid, ACS Grade; 1 L
8002952

$250.48CAD / Each
+ +
LabChem Potassium Iodide, 10% (w/v); 500 mL
8003513

$103.73CAD / Each
+ +
LabChem Sodium Hydroxide, 10% (w/w); 1 L
8003839

$70.28CAD / Each
=
Total Price 1184.92 ADD 10 ITEMS TO CART
Product Description

Key Features

  • Obtain quick ion concentration measurements in the field or laboratory
  • Dip and compare color change — no calibration, electricity, or warm up needed

More About this Item

These ion test strips make water testing fast and easy. Conveniently packed in screw top bottles, the test papers are safe even in rugged field applications. Obtaining simultanious pH and ion concentration results couldn't be easier. Simply dip the test strip in your water sample, wait about 15 seconds for the water to penetrate the strip, and compare the color changes against the chart on the bottle's label.

Specifications
  • TestCyanogen
  • Measurement Points0 to 1000 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionTest Strips, Cyanogen Check (Cyanide Cn-), 0 to 1000 ppm; 100 Strips/Pk
Reviews
Q&A