Advantec pH Test Papers and Indicator Strips
Determine pH easily and inexpensively without instrumentation
  • Color changes instantaneously, simply compare to chart for determination
  • Available in two convenient formats — a booklet of 20 strips, 7 x 70 mm; or a single 9-mm wide roll in a convenient dispenser
$27.41 - $54.52CAD / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Measurement Points
Availability Pricing
Advantec Litmus Blue Test Paper, 10 Booklets of 20 Strips 5920100 Mfr # 07020010
COMPARE
Presence/absence
$27.41CAD / Pkg of 10
Advantec Litmus Red Test Paper, 10 Booklets of 20 Strips 5920102 Mfr # 07020020
COMPARE
Presence/absence
$27.41CAD / Pkg of 10
Advantec Universal pH Test Paper; pH Range 1 to 11; 10 Booklets of 20 Strips 5920104 Mfr # UNIV-B
COMPARE
1 to 11 pH
$44.50CAD / Pkg of 10
Advantec UNIV-R Universal pH Test Paper; pH Range 1 to 11; 9 mm x 6 m Roll 5920106 Mfr # UNIV-R
COMPARE
1 to 11 pH
$54.52CAD / Pkg of 1
Advantec W-R Whole Range pH Test Paper; pH Range 1 to 14; 9 mm x 5.5 m Roll 5920108 Mfr # W-R
COMPARE
1 to 14 pH
$45.38CAD / Pkg of 1