Product Type
In Stock
Industry
Capacity (Liters)
Power (VAC)
Capacity (cu ft)
Regulatory Compliance
Max Temperature (° C)
Control Type
CO2 Incubators
CO2 Incubators
CO2 incubators are used for conducting cell culture and tissue culture applications. CO2 incubators are available with either thermal conductivity (TC) or infrared (IR) CO2 sensors. Both respond quickly to changes in CO2 levels and help provide excellent CO2 control. The TC sensor is recommended for application in which chamber temperature and humidity are relatively constant; the IR sensor is more suitable for applications where temperature and humidity levels are frequently changing.
APPLY FILTERS
filter byclear all
4 RESULTS
SORT BY
Showing 1 - 4 of 4