Water Purification
Water Purification
Methods of Purification    Methods of Purification Table     ASTM Water Grade Standards
Methods of Purification
Deionization
The removal of ions and minerals by synthetic ion exchange resins. Cation resins remove positively charged ions; anion resins remove negatively charged ions.
Adsorption
Activated carbon bonds with the chlorine and organic materials in feedwater to immobilize and remove these impurities.
Filtration
Use as a pretreatment or a stand-alone treatment. Water passes through a filter of specified porosity at normal line pressures. The filter retains most particulates, with water passing through.
Ultrafiltration
Use for removing pyrogens and bacteria. Under pressure, water is forced through a membrane with a pore size smaller than 0.005 µm. Particulates are retained, with only pure water passing through.
Reverse Osmosis
Use primarily as a pretreatment. Equal amounts of pure water and saline solution are separated in a U-tube by a semipermeable membrane. When external pressure is applied to the saline side, the semipermeable membrane allows water to pass through while salts are concentrated and flushed down a drain.
Distillation
The process in which water is heated to a gaseous state and recondensed in a separate vessel.
Ultraviolet (UV) Oxidation
UV light (at <280 nm) passes through the water destroying bacteria, viruses, and trace organics.
Methods of Purification Table
Method Dissolved
ionized solids
Dissolved
organics
Dissolved
ionized gases
Particulates Bacteria Pyrogens
Deionization Excellent Poor Excellent Poor Poor Poor
Adsorption Poor Excellent Poor Poor Poor Poor
Filtration Poor Poor Poor Excellent Excellent Poor
Ultrafiltration Poor Good Poor Excellent Excellent Excellent
Reverse osmosis Good Good Poor Excellent Excellent Excellent
Distillation Excellent
/Good
Good Poor Excellent Excellent Excellent
UV oxidation Poor Good Poor Poor Good Poor
ASTM Water Grade Standards
Resistivity
(MΩ-cm, min)
Conductivity
(µS/cm, max)
Organic carbon
(µg/L, max)
Silica
(µg/L, max)
Chlorides
(µg/L, max)
Sodium
(µg/L, max)
Type I 18.0 0.056 100 3 1 1
Type II 1.0 1.0 50 3 5 5
Type III 4.0 0.25 200 500 10 10
Type IV 0.2 5.0 No limit No limit 50 50