• Cole-Parmer
  • Environmental Express
  • ZeptoMetrix
  • ;
  • Sign In
    • Log in for these benefits:
    • View Online Orders Status View Online Orders Status
    • Review Order History Review Order History
    • Create Online Quotes Create Online Quotes
    Sign In
    New Customer? Register Now
  • Request A Quote
  • ORDER STATUS
  • Shop Products
  • Canada
  • Help
  • Search
Geust User Link
Cole-Parmer
  • Quick Order
×

Enter Each Item Number & Quantity

Please see error(s) highlighted below.
Add more products on the full Quick Order page
Add to Cart Cancel
×
  • Sign Into

    My Account

      • Log in for these benefits:
      • View Online Orders Status View Online Orders Status
      • Review Order History Review Order History
      • Create Online Quotes Create Online Quotes
    • Sign In
      New Customer? Register Now
  • Canada
  • Products

    Popular Categories

    Laboratory Equipment Laboratory Equipment
    Data Collection Products Data Collection Products
    Safety Products Safety Products
    Balances and Scales Balances and Scales
    see all categories
    • Balances and Scales
      • Analytical Balances
      • Bench and Floor Scale Bases
      • Bench and Floor Scales
      • Counting Scales
      • Crane and Hanging Scales
      • Health Scales
      • Indicators for Scales
      • Mechanical Balances
      • Moisture Balances
      • Platform and Pallet Scales
      • Production Weighing
      • Semi and Micro Analytical Balances
      • Toploading Balances
      • Weighing Accessories
      • Weighing Dishes
    • Calibration
      • Calibration Accessories
      • Calibration Equipment
      • Calibration Masses
      • Calibration Services
    • Electrical Products
      • Batteries and Battery Testers
      • Cable Testers
      • Cable Ties
      • Clamp Meters
      • Electrical Accessories
      • Electronics Enclosures
      • Function Generators
      • Gauss Meters
      • Insulation Testers
      • LCR Meters
      • Megohmmeters
      • Motor Rotation Testers
      • Multimeters
      • Oscilloscopes
      • Outlet Strips
      • Power Meters and Analyzers
      • Power Supplies
      • Resistance Testers
      • RF Signal Generators
      • Static Control Products
      • Surge Protectors
      • Test Leads and Probes
      • Voltage Detectors
    • Filtration Products
      • Laboratory Filtration Products
      • Process Filtration Products
    • Fluid Handling
      • Fittings
      • Flowmeters
      • Sensors and Monitors
      • Tubing and Hose
      • Valves
    • Furniture
      • Antivibration Platforms and Tables
      • Benches and Tables
      • Chairs and Stools
      • Cleanroom Furniture
      • Fume Hoods
      • Furniture Accessories
      • Glove Boxes
      • PCR Chambers
      • Shelving
      • Storage Cabinets
      • Workstations
    • Heating and Cooling Equipment
      • Cryogenic Products
      • Environmental Chambers
      • Heaters
      • Ice Machines
      • Incubators
      • Lab Burner
      • Laboratory Furnaces
      • Ovens
      • Refrigerators and Freezers
      • Steam Generators
      • Sterilizers
    • Laboratory Equipment
      • Baths and Circulators
      • Burettes (Burets)
      • Calorimeters
      • Centrifuges
      • Chromatography Products
      • Desiccators
      • Digestion Systems
      • Dispensers
      • Distillation
      • Electrophoresis Products
      • Electroporation Products
      • Environmental Samplers
      • Extraction Apparatus
      • Fermentation Products
      • Flame Photometers
      • Fluid Bed Dryers
      • Freeze Dryers
      • Glassware Washers
      • Histology Products
      • Homogenizers and Ultrasonic Processors
      • Humidifiers and Dehumidifiers
      • Lab Shakers
      • Laboratory Evaporators
      • Laboratory Sieves and Sieve Shakers
      • Leachate Equipment
      • Lighting and Illumination
      • Magnetic Stirrers and Hot Plates
      • Microscopes and Magnification
      • Mixers and Blenders
      • Particle Counters
      • Pipettes and Pipettors
      • Refractometers
      • Thermal Cyclers
      • Titrators
      • Ultrasonic Cleaners
      • Vacuums
      • Water Purification Products
    • Laboratory Supplies
      • Air Sampling
      • Animal Research Products
      • Bags
      • Beakers
      • Books and Reference Materials
      • Bottles and Jars
      • Calculators
      • Carboys
      • Chemistry Kits
      • Clean Room Stationery
      • Containers
      • Cuvettes
      • Dialysis Products
      • Dippers
      • Distillation Supplies
      • Field Sampling
      • Flasks
      • Graduated Cylinders
      • Ice Buckets and Ice Pans
      • Lab Cleaning Products
      • Lab Organizers
      • Labels and Label Printers
      • Laboratory Dishes
      • Laboratory Timers and Clocks
      • Labware
      • Microbiology Products
      • Microplate Products
      • Mortars and Pestles
      • Parts and Accessories
      • Racks
      • Sampling Wipes
      • Shippers
      • Syringes
      • Tubes
      • Vials
      • Watch Glasses
    • Material Handling and Storage
      • Carts
      • Hand and Platform Trucks
      • Stools and Ladders
      • Storage and Handling
      • Tanks and Drums
      • Tools
      • Worktables and Trucks
    • Material Testing and Physical Testing
      • Color and Gloss Meters
      • Density and Specific Gravity
      • Force and Torque
      • Hardness Testers
      • Melting Point Apparatuses
      • Petroleum Testing Products
      • Polarimeters
      • Presses
      • Spectroscopy and Spectrophotometers
      • Surface Roughness Testers
      • Tensiometers
      • Thickness Gauges
      • Viscometers
      • Viscosity Cups
    • Pressure and Vacuum Instruments
      • Gauge guards, Seals, and Accessories
      • Pressure Calibrators
      • Pressure Gauges
      • Pressure I/P Transducers
      • Pressure Regulators
      • Pressure Switches
      • Pressure Transducers
      • Vacuum Controllers, Gauges, and Systems
    • Process Controllers
      • Automation Control
      • Conductivity, Resistivity, and TDS Control
      • Dissolved Oxygen Control
      • Ion Control
      • Multiparameter Control
      • Panel Meters
      • pH/ORP Control
      • Temperature Process Controllers and Alarms
    • Pumps
      • Air Compressors
      • Drum and Hand Pumps
      • Peristaltic Pumps
      • Piston Pumps
      • Positive Displacement Pumps
      • Pump Parts and Accessories
      • Syringe Pumps
      • Transfer Pumps
      • Vacuum Pumps
    • Safety Products
      • Air Quality Monitors
      • Arc Flash Products
      • Combustion Analyzers
      • Eyewash
      • Eyewear and Safety Glasses
      • Floor Mats and Matting
      • Gas Detectors
      • Gloves
      • Hearing Protection
      • Lockout and Tagout Product
      • Personal Monitor Badges
      • Radiation Detectors
      • Refrigerant Products
      • Respirators and Masks
      • Safety Apparel
      • Safety Shields
      • Safety Signs and Labels
      • Solvent Safety
      • Sound Meters
      • Spill Control
    • Temperature Measurement Products
      • Bimetal Thermometers
      • Digital Thermometers
      • Glass Thermometers
      • Infrared Thermometers
      • RTD Thermometers
      • Temperature Accessories
      • Temperature Controllers
      • Temperature Labels
      • Temperature Probe Accessories
      • Temperature Probes
      • Temperature Switches
      • Thermal Imagers
      • Thermistor Meters
      • Thermocouple Meters
    • Test and Measurement Products
      • Airflow Measurement Products
      • Data Collection Products
      • Humidity Products
      • Hydrometers
      • Light Meters
      • Moisture Determination Products
      • Moisture Meters
      • Penetrometers
      • Stroboscopes
      • Tachometers
      • Vibration Measurement
      • Weather Measurement Products
    • Water Quality Products
      • Colorimeters
      • Colorimetry Reagents
      • Conductivity, Resistivity, and TDS Meters
      • Dissolved Oxygen Meters
      • Dye Tracers
      • Electrodes
      • Flocculation Testers
      • Multiparameter Meters
      • pH Buffers
      • pH Meters
      • Turbidity Accessories
      • Turbidity Meters
      • Water Quality Accessories
      • Water Quality Standards, and Solutions
      • Water Quality Test Strips

    Featured Links

    • All Categories A-Z
    • Always In Stock
    • Best Sellers
    • Buy More And Save
    • Specials And Promotions
    • Specialty Brands
    • What's New
    CDC Compliant Virus Testing Products
  • Products
    Piston Pumps
    Tubing and Hose
    Laboratory Equipment
    Water Quality Products
    Test and Measurement Products
    < Back
    • Fluid Handling
      • Fittings
      • Flowmeters
      • Sensors and Monitors
      • Tubing and Hose
      • Valves
    • Laboratory Equipment
      • Baths and Circulators
      • Burettes (Burets)
      • Calorimeters
      • Centrifuges
      • Chromatography Products
      • Desiccators
      • Digestion Systems
      • Dispensers
      • Distillation
      • Electrophoresis Products
      • Electroporation Products
      • Environmental Samplers
      • Extraction Apparatus
      • Fermentation Products
      • Flame Photometers
      • Fluid Bed Dryers
      • Freeze Dryers
      • Glassware Washers
      • Histology Products
      • Homogenizers and Ultrasonic Processors
      • Humidifiers and Dehumidifiers
      • Lab Shakers
      • Laboratory Evaporators
      • Laboratory Sieves and Sieve Shakers
      • Leachate Equipment
      • Lighting and Illumination
      • Magnetic Stirrers and Hot Plates
      • Microscopes and Magnification
      • Mixers and Blenders
      • Particle Counters
      • Pipettes and Pipettors
      • Refractometers
      • Thermal Cyclers
      • Titrators
      • Ultrasonic Cleaners
      • Vacuums
      • Water Purification Products
    • Pumps
      • Air Compressors
      • Drum and Hand Pumps
      • Peristaltic Pumps
      • Piston Pumps
      • Positive Displacement Pumps
      • Pump Parts and Accessories
      • Syringe Pumps
      • Transfer Pumps
      • Vacuum Pumps
    • Process Controllers
      • Automation Control
      • Conductivity, Resistivity, and TDS Control
      • Dissolved Oxygen Control
      • Ion Control
      • Multiparameter Control
      • Panel Meters
      • pH/ORP Control
      • Temperature Process Controllers and Alarms
    • Pressure and Vacuum Instruments
      • Gauge guards, Seals, and Accessories
      • Pressure Calibrators
      • Pressure Gauges
      • Pressure I/P Transducers
      • Pressure Regulators
      • Pressure Switches
      • Pressure Transducers
      • Vacuum Controllers, Gauges, and Systems
    • Water Quality Products
      • Colorimeters
      • Colorimetry Reagents
      • Conductivity, Resistivity, and TDS Meters
      • Dissolved Oxygen Meters
      • Dye Tracers
      • Electrodes
      • Flocculation Testers
      • Multiparameter Meters
      • pH Buffers
      • pH Meters
      • Turbidity Accessories
      • Turbidity Meters
      • Water Quality Accessories
      • Water Quality Standards, and Solutions
      • Water Quality Test Strips
    • Test and Measurement Products
      • Airflow Measurement Products
      • Data Collection Products
      • Humidity Products
      • Hydrometers
      • Light Meters
      • Moisture Determination Products
      • Moisture Meters
      • Penetrometers
      • Stroboscopes
      • Tachometers
      • Vibration Measurement
      • Weather Measurement Products
    • Temperature Measurement Products
      • Bimetal Thermometers
      • Digital Thermometers
      • Glass Thermometers
      • Infrared Thermometers
      • RTD Thermometers
      • Temperature Accessories
      • Temperature Controllers
      • Temperature Labels
      • Temperature Probe Accessories
      • Temperature Probes
      • Temperature Switches
      • Thermal Imagers
      • Thermistor Meters
      • Thermocouple Meters
    • Safety Products
      • Air Quality Monitors
      • Arc Flash Products
      • Combustion Analyzers
      • Eyewash
      • Eyewear and Safety Glasses
      • Floor Mats and Matting
      • Gas Detectors
      • Gloves
      • Hearing Protection
      • Lockout and Tagout Product
      • Personal Monitor Badges
      • Radiation Detectors
      • Refrigerant Products
      • Respirators and Masks
      • Safety Apparel
      • Safety Shields
      • Safety Signs and Labels
      • Solvent Safety
      • Sound Meters
      • Spill Control
    • Laboratory Supplies
      • Air Sampling
      • Animal Research Products
      • Bags
      • Beakers
      • Books and Reference Materials
      • Bottles and Jars
      • Calculators
      • Carboys
      • Chemistry Kits
      • Clean Room Stationery
      • Containers
      • Cuvettes
      • Dialysis Products
      • Dippers
      • Distillation Supplies
      • Field Sampling
      • Flasks
      • Graduated Cylinders
      • Ice Buckets and Ice Pans
      • Lab Cleaning Products
      • Lab Organizers
      • Labels and Label Printers
      • Laboratory Dishes
      • Laboratory Timers and Clocks
      • Labware
      • Microbiology Products
      • Microplate Products
      • Mortars and Pestles
      • Parts and Accessories
      • Racks
      • Sampling Wipes
      • Shippers
      • Syringes
      • Tubes
      • Vials
      • Watch Glasses
    • Material Testing and Physical Testing
      • Color and Gloss Meters
      • Density and Specific Gravity
      • Force and Torque
      • Hardness Testers
      • Melting Point Apparatuses
      • Petroleum Testing Products
      • Polarimeters
      • Presses
      • Spectroscopy and Spectrophotometers
      • Surface Roughness Testers
      • Tensiometers
      • Thickness Gauges
      • Viscometers
      • Viscosity Cups
    • Material Handling and Storage
      • Carts
      • Hand and Platform Trucks
      • Stools and Ladders
      • Storage and Handling
      • Tanks and Drums
      • Tools
      • Worktables and Trucks
    • Heating and Cooling Equipment
      • Cryogenic Products
      • Environmental Chambers
      • Heaters
      • Ice Machines
      • Incubators
      • Lab Burner
      • Laboratory Furnaces
      • Ovens
      • Refrigerators and Freezers
      • Steam Generators
      • Sterilizers
    • Electrical Products
      • Batteries and Battery Testers
      • Cable Testers
      • Cable Ties
      • Clamp Meters
      • Electrical Accessories
      • Electronics Enclosures
      • Function Generators
      • Gauss Meters
      • Insulation Testers
      • LCR Meters
      • Megohmmeters
      • Motor Rotation Testers
      • Multimeters
      • Oscilloscopes
      • Outlet Strips
      • Power Meters and Analyzers
      • Power Supplies
      • Resistance Testers
      • RF Signal Generators
      • Static Control Products
      • Surge Protectors
      • Test Leads and Probes
      • Voltage Detectors
    • Calibration
      • Calibration Accessories
      • Calibration Equipment
      • Calibration Masses
      • Calibration Services
    • Balances and Scales
      • Analytical Balances
      • Bench and Floor Scale Bases
      • Bench and Floor Scales
      • Counting Scales
      • Crane and Hanging Scales
      • Health Scales
      • Indicators for Scales
      • Mechanical Balances
      • Moisture Balances
      • Platform and Pallet Scales
      • Production Weighing
      • Semi and Micro Analytical Balances
      • Toploading Balances
      • Weighing Accessories
      • Weighing Dishes
    • Filtration Products
      • Laboratory Filtration Products
      • Process Filtration Products
    • Furniture
      • Antivibration Platforms and Tables
      • Benches and Tables
      • Chairs and Stools
      • Cleanroom Furniture
      • Fume Hoods
      • Furniture Accessories
      • Glove Boxes
      • PCR Chambers
      • Shelving
      • Storage Cabinets
      • Workstations
  • Industries & Workflows

    Featured Industries

    • Botanicals
    • Cold Chain
    • Environmental
    • Pharmaceutical
    • Vaccine Research and Development
    • Virus Testing
    • Wastewater
    view all industries & workflows
    • Sample Preparation for Lab Analysis
    • Lab Milling and Grinding
    • Lab Mixing and Blending
    • Tissue Homogenization and Cell Lysis
  • Brands
    Featured Brands
    • Cole-Parmer
    • Environmental Express Oakton
    • Traceable
    • Digi-Sense
    view all brands
    Traceable Products Calibrated Monitoring
    Vaplock closed systems for solvent containment made easy
  • Services
    • Calibration Services by InnoCal
    • Custom Temperature Probes
    • Help
    view all services
  • Product Support
    Technical Resource Library
    • Technical Resource Library
    • Articles and White Papers
    • Videos
    • Webinars
    • Technical Resource Map
    Tools & Compatibility
    • Chemical Compatibility Database
    • Tubing Chemical Compatibility
    • Tubing and Food Compatibility
    • Tubing Selector Tool
    • Common Unit Converters
    • Safety Glove Chemical Compatibility Database
    More Product Support
    • Certificate Lookup
    • Downloadable Literature
    • Regulatory Agencies
    • Technical Glossary
    View All Resources
  • Home
  • Technical Resource Library
  • Videos
+ SAVE TO MY DOCS
  • Print

AVAILABLE IN: EnglishFrench
Last Updated: 05/03/21

Sign up to receive the latest in industry news, products, & services.

You have successfully subscribed to our newsletter
  • Facebook
  • LinkedIn
  • Twitter
  • Instagram
  • YouTube
Since 1955
Meeting the needs of the scientific and engineering communities since 1955.
Company Info
  • About Us
  • Quality Policy
  • Careers
  • Testimonials
  • Equal Employment Opportunity
  • Business Profile
  • Press Releases
  • Event Calendar
  • Cole-Parmer Blog
Order Support
  • Request a Quote
  • Order Status
  • Quote Status
  • Returns & Cancellations
  • Shipping Policy
  • Warranty Policy
  • Terms & Conditions
Customer Support
  • Contact Us
  • Cookie Policy
  • Locate a Dealer
  • Site Support
  • FAQs
© 2026 Cole-Parmer Instrument Company, LLC. All Rights Reserved.
  • Terms of Use
  • Privacy Policy
  • Site Map
go BACK TO TOP