Selecting the right syringe pump is essential for ensuring precision and reliability in fluid handling applications. Cole-Parmer offers a broad range of KD Scientific syringe pumps tailored to meet laboratories' evolving needs, from simple infusion to sophisticated multi-step protocols. This article compares the Legacy KDS 100, Legato® 100, and Legato® 200 series syringe pumps, outlining the key differences, ideal applications, and tips for getting the most out of your system.
KD Scientific Pump Series Overview
Legacy Series KDS 100
The Legacy KDS 100 is the foundation of KD Scientific’s long-standing reputation for combining precision and simplicity. It offers reliable infusion functionality with essential features, including a backlit LCD for clear visibility and an intuitive setup process. Users can easily configure the flow rate, select syringe size, and set dispense volume, making it simple to tailor the pump to specific application needs. Volume and flow rate units (µL/hr or mL/hr) are selected automatically based on syringe size. The KDS 100 remains a dependable and cost-effective choice for straightforward infusion applications where simplicity and reliability are top priorities.
Legato Series
The Legato series represents the next generation of syringe pump innovation, combining superior flow performance with intuitive usability and advanced connectivity. The high-resolution 4.3" color touchscreen display is a standout feature, which shows all pump operating parameters on a single screen including, flow rate, volume delivered, elapsed time, time remaining, and syringe selection. Core features like USB, TTL, and RS-485 communication enable seamless integration with PCs and laboratory automation systems. Daisy-chain up to 99 pumps for synchronized multi-pump workflows. Unique to the Legato design, the screen orientation rotates automatically when placed on its side, reducing the pump’s footprint by four times to maximize valuable bench space. The adjustable linear force mechanism ensures optimal pressure is applied to a wide range of syringe sizes, while the rugged welded steel chassis minimizes vibration and noise, even during high-pressure operation enhancing stability and performance. The Legato 100 series delivers these essential capabilities in a cost-effective platform ideal for single- or dual-syringe infusion and infuse/withdraw applications with excellent precision and flow stability.
Engineered for performance and flexibility, the Legato 200 series builds on the capabilities of the Legato 100, making it ideal for demanding research and industrial environments. The 200 series accommodates larger syringe volumes and includes RS-232 communication for expanded system integration. With higher linear force, these pumps are well-suited for viscous fluids and high-pressure applications. Integrated features like calibration reminders, dual external triggers, and a real-time clock display make this series a go-to choice for complex, high-precision workflows.
How to Choose the Right Pump
Selecting the right syringe pump starts with identifying your application's key requirements. Considering these factors will help narrow down your options and ensure optimal performance and usability.
Number of Syringes
- Single syringe pumps are ideal for simple tasks where only one reagent or fluid needs to be handled at a time.
- Dual syringe pumps allow simultaneous or alternating functions, enabling continuous flow or precise volume matching.
Programmable
- Basic control with no programming offers straightforward operation, making them ideal for fixed-rate applications where parameters remain consistent throughout the process.
- Limited programming allows users to create and store two custom programs with up to 50 steps each, providing flexibility for simple, repeatable workflows.
- Advanced programmability offers the most robust programming options, including a single complex program with up to 100 steps or storage for 40 programs with 25 steps each. This is ideal for high-precision, multi-phase protocols or regulated environments requiring method validation.
Syringe Size
- Smaller syringes provide higher precision for nanoliter/picoliter flows, while larger syringes support high-volume transfers and longer run times.
- Use the smallest viable syringe size for higher flow control resolution.
Cole-Parmer’s Syringe Pump Selection
Cole-Parmer offers a comprehensive selection of KD Scientific syringe pumps. Utilize these factors to match your application needs with the right model for optimal performance, accuracy, and workflow efficiency.
| Item Number | Model | # of Syringes | Mode | Programmable | Syringe Size | Flow Rate | Voltage |
| 51005-10 | Legacy KDS 100 | 1 | Infusion | No | 10 µL to 60 mL | 1667 pL/min to 7.05 mL/min | 110 VAC |
| 51005-11 | Legacy KDS 100 | Infusion | No | 10 µL to 60 mL | 1667 pL/min to 7.05 mL/min | 230 VAC |
| 51005-01 | Legato 100 | Infusion | No | 0.5 µL to 60 mL | 1.28 pL/min to 88.28 mL/min | 100–240 VAC |
| 51005-03 | Legato 110 | Infusion/Withdrawal | 2 Programs/50 steps each | 0.5 µL to 60 mL | 1.28 pL/min to 88.28 mL/min | 100–240 VAC |
| 51005-06 | Legato 185 | Infusion/Withdrawal | 2 Programs/50 steps each | 0.5 µL to 60 mL | 0.54 pL/min to 39 mL/min | 100–240 VAC |
| 51005-02 | Legato 101 | 2 | Infusion | No | 0.5 µL to 10 mL | 1.28 pL/min to 25.99 mL/min | 100–240 VAC |
| 51005-04 | Legato 111 | Infusion/Withdrawal | 2 Programs/50 steps each | 0.5 µL to 10 mL | 1.28 pL/min to 25.99 mL/min | 100–240 VAC |
| 51005-05 | Legato 180 | Infusion/Withdrawal | 2 Programs/50 steps each | 0.5 µL to 10 mL | 0.58 pL/min to 11.70 mL/min | 100–240 VAC |
| 51005-07 | Legato 200 | Infusion | No | 0.5 µL to 140 mL | 3.06 pL/min to 215.8 mL/min | 100–240 VAC |
| 51005-08 | Legato 210 | Infusion/Withdrawal | 1 Program/100 steps | 0.5 µL to 140 mL | 3.06 pL/min to 215.8 mL/min | 100–240 VAC |
| 51005-09 | Legato 210P | Infusion/Withdrawal | 40 Programs/25 steps | 0.5 µL to 140 mL | 3.06 pL/min to 215.8 mL/min | 100–240 VAC |
Essential Supporting Components
For optimal performance and compatibility, consider these accessories:
- Syringes: Choose from gas-tight, glass, or stainless steel syringes depending on your chemical compatibility and pump capabilities.
- Luer Fittings: Ensure secure, leak-proof connections between syringes and your downstream fluid path. Use a luer lock to hose barb fittings to ensure a tight, leak-free connection to flexible tubing, especially under pressure or during extended runs.
- Tubing: Select tubing based on the chemical compatibility of the fluid being pumped to ensure material integrity and avoid contamination. Also consider application-specific certifications such as USP Class VI, FDA, or ISO for regulated environments to ensure compliance and system safety.
Real-World Applications and Recommended Models
| Application | Ideal Pump | Reason |
| Electrospinning or MS Calibration | Legacy KDS 100 or Legato 100 | Basic infusion functionality with reliable flow and easy setup. |
| Surface Plasma Resonance, Microdialysis | Legato 101 | Dual syringe configuration and nanoliter flow precision for low-volume applications. |
| Dosing or Nutrient Delivery | Legato 110 or 111 | Infuse/withdraw modes with programmable steps for precise dosing control. |
| Microfluidics, Ultra-Low-Flow Delivery | Legato 180 | Picoliter flow rates with high reproducibility for exact applications. |
| Cell Injections, Neuroscience | Legato 185 | Single-syringe picoliter delivery for delicate, high-precision dosing. |
| Drug Discovery, Flow Chemistry | Legato 200 | Supports larger syringe volumes and viscous fluids. |
| Biopharma R&D | Legato 210 | Advanced control, multi-mode operation, and compliance-friendly features. |
| Organic Synthesis, Complex Dosing | Legato 210 or 210P | Multi-step programmability and event-triggered operations for automated workflows. |
Tips for Long-Term and Optimal Use
- Routine Calibration: Legato 200 models feature reminders and diagnostics for timely calibration and maintenance.
- Syringe Accuracy: Measure the syringe bore diameter to enter precise syringe dimensions. Use the entry option to input the actual value in millimeters (mm) to optimize flow rate calculations.
- Mechanical Maintenance: Regularly inspect and lubricate the guide rods and lead screw to maintain smooth motion and extend pump life.
- Linear Force Adjustment: Set the appropriate linear force based on syringe size and fluid viscosity to prevent stalling or syringe damage.
Select with Confidence
From reliable infusions to complex operations, KD Scientific pumps offered by Cole-Parmer support a wide range of applications in the lab and beyond.
Explore Cole-Parmer’s complete syringe pump portfolio or contact our technical specialists to find the best solution for your application needs.