Absolute, Gauge, and Differential Pressure Scales
Absolute, Gauge, and Differential Pressure Scales
Use differential pressure sensors to measure gauge pressure by leaving one process connection open to atmosphere and connecting the second sensor port to your system.