W.S. Tyler Ro-tap II Shakers for 8" Diameter Sieves
Reproduce the rotating and tapping motion of hand-sieving but with more uniform motion for highly reproducible results
Shakers are actuated by a built-in, 99-minute digital timer with 0.1 second accuracy. They are powered by a 1/4 hp motor and include a 5-ft cord (110 VAC models also include U.S. three-prong plug).
Ro-tap II Sieve Shakers double your shaking capacity while maintaining the accuracy of a single mechanical shaker. Units hold up to twelve full-height sieves or twenty-six half-height sieves.
$9,967.48 - $11,647.15CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) RPM Timer Range (min)
Availability Pricing
W.S. Tyler Ro-Tap II Sieve Shaker for 8" diameter sieves, 110 VAC, 60 Hz 5998650 Mfr # RX-94
COMPARE
110 1750 99
$11,647.15CAD / Each
W.S. Tyler Ro-Tap II Sieve Shaker for 8" diameter sieves, 220 VAC, 50 Hz 5998652 Mfr # RX-94-3
COMPARE
220 1450 99
$9,967.48CAD / Each