VistaLab™ Stacked Rack Non-Sterile Pipette Tips
Maximize performance and benefits of your pipette
  • Tips made of 100% pure virgin polypropylene
These eco-friendly polypropylene (PP) stacked rack pipette tips are packaged as 200 tips per rack and are available in 3 and 5 rack packages. 250 and 1250 µL tips are compatible with VistaLab™ Ovation and MLA model pipettors. 25 µL tips are only compatible with VistaLab™ Ovation model pipettors.
$108.00 - $131.28CAD / Pkg of 1000
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Package Of Tip Style
Availability Pricing
VistaLab Stacked Rack Pipette Tips, Non-Sterile, 250 µL; 200 Tips x 5 Racks 0619991 Mfr # 9025
COMPARE
250 Racks; 1000/Pk Small; Standard
$131.28CAD / Pkg of 1000
VistaLab Stacked Rack Pipette Tips, Non-Sterile, 1250 µL; 200 Tips x 3 Racks 0619993 Mfr # 9026
COMPARE
1250 Racks; 600/Pk Large; Standard
$108.00CAD / Pkg of 600