Versilon® SPT-50 Platinum-Cured Silicone Tubing
Superior inner surface smoothness and minimal extractables—perfect for transferring high-purity fluids
  • Excellent bend radius and flexibility
  • Does not transmit taste or odor
  • High tear resistance leads to long service life
  • Great stability over a wide temperature range
  • Translucent tubing provides good clarity
  • Manufactured and packaged in a cleanroom environment
  • Sterilize by autoclave, gamma irradiation, or EtO
  • Complies with FDA 21 CFR 177.2600 and NSF-51
  • Ideal for food and beverage, general-purpose, and laboratory applications
  • Free technical application support available to assist with tubing selection
The Versilon® SPT-50 platinum-cured silicone tubing features exceptionally smooth inner surface, minimal extractables, and does not transmit taste or odors, making it perfect for transferring high-purity fluids. The smooth bore inhibits particle entrapment, protein binding, and bacterial growth, ensuring the integrity of the fluid. Excellent flexibility, high tear resistance, and non-tacky outer surface make this tubing ideal for peristaltic pump applications.
$581.99 - $2,386.89CAD / Pkg of 1
32 variations of this product are available.
See All Product Options Below
32 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Versilon SPT-50 Platinum-Cured Silicone Tubing, 1/32" ID x 3/32" OD; 50 Ft 0012338 Mfr # SPT-50 Tubing - .031 X .094
COMPARE
1/32 3/32 1/32
$581.99CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 1/16" ID x 1/8" OD; 50 Ft 0012339 Mfr # SPT-50 Tubing - .063 X .125
COMPARE
1/16 1/8 1/32
$581.99CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 1/16" ID x 3/16" OD; 50 Ft 0012340 Mfr # SPT-50 Tubing - .063 X .188
COMPARE
1/16 3/16 1/16
$604.09CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 3/32" ID x 5/32" OD; 50 Ft 0012341 Mfr # SPT-50 Tubing - .094 X .156
COMPARE
3/32 5/32 1/32
$581.99CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 3/32" ID x 7/32" OD; 50 Ft 0012342 Mfr # SPT-50 Tubing - .094 X .219
COMPARE
3/32 7/32 1/16
$604.09CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 1/8" ID x 3/16" OD; 50 Ft 0012343 Mfr # SPT-50 Tubing - .125 X .188
COMPARE
1/8 3/16 1/32
$581.99CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 1/8" ID x 1/4" OD; 50 Ft 0012344 Mfr # SPT-50 Tubing - .125 X .250
COMPARE
1/8 1/4 1/16
$618.82CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 5/32" ID x 7/32" OD; 50 Ft 0012345 Mfr # SPT-50 Tubing - .156 X .219
COMPARE
5/32 7/32 1/32
$604.09CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 3/16" ID x 1/4" OD; 50 Ft 0012346 Mfr # SPT-50 Tubing - .187 X .250
COMPARE
3/16 1/4 1/32
$604.09CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 3/16" ID x 5/16" OD; 50 Ft 0012347 Mfr # SPT-50 Tubing - .187 X .313
COMPARE
3/16 5/16 1/16
$618.82CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 3/16" ID x 3/8" OD; 50 Ft 0012348 Mfr # SPT-50 Tubing - .187 X .375
COMPARE
3/16 3/8 3/32
$648.29CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 3/16" ID x 7/16" OD; 50 Ft 0012349 Mfr # SPT-50 Tubing - .187 X .438
COMPARE
3/16 7/16 1/8
$699.86CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 1/4" ID x 5/16" OD; 50 Ft 0012350 Mfr # SPT-50 Tubing - .250 X .313
COMPARE
1/4 5/16 1/32
$604.09CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 1/4" ID x 3/8" OD; 50 Ft 0012351 Mfr # SPT-50 Tubing - .250 X .375
COMPARE
1/4 3/8 1/16
$633.56CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 1/4" ID x 7/16" OD; 50 Ft 0012352 Mfr # SPT-50 Tubing - .250 X .438
COMPARE
1/4 7/16 3/32
$685.13CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 1/4" ID x 1/2" OD; 50 Ft 0012353 Mfr # SPT-50 Tubing - .250 X .500
COMPARE
1/4 1/2 1/8
$721.96CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 5/16" ID x 7/16" OD; 50 Ft 0012354 Mfr # SPT-50 Tubing - .312 X .438
COMPARE
5/16 7/16 1/16
$648.29CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 5/16" ID x 1/2" OD; 50 Ft 0012355 Mfr # SPT-50 Tubing - .312 X .500
COMPARE
5/16 1/2 3/32
$699.86CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 3/8" ID x 1/2" OD; 50 Ft 0012356 Mfr # SPT-50 Tubing - .375 X .500
COMPARE
3/8 1/2 1/16
$670.39CAD / Pkg of 1
Versilon SPT-50 Platinum-Cured Silicone Tubing, 3/8" ID x 9/16" OD; 50 Ft 0012357 Mfr # SPT-50 Tubing - .375 X .563
COMPARE
3/8 9/16 3/32
$721.96CAD / Pkg of 1
Showing 1 - 20 of 32