Versilon® Silver Antimicrobial Tubing

Silver-based antimicrobial compound inhibits growth of microbes
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • Plasticizer-free inner bore—excellent for food and beverage applications
  • Reduction in microbe growth effectively minimizes foul odors and discoloration
  • Meets FDA criteria
  • Phthalate and BPA free
Formulated with silver-based antimicrobial compound on the inner diameter surface, Versilon® Silver tubing offers excellent protection against foul odors, discoloration, and formation of mildew and biofilm. Not only do microbes degrade tubing, but it can also contaminate the material being transferred. While tubing can be washed with detergent and hot water to kill the microbes, this can be time consuming and costly, and it does not provide residual protection against fresh contamination. With the antimicrobial compound embedded in the inner surface, Versilon Silver tubing ensures the integrity of the fluid by providing lasting protection against microbe growth.
$216.59 - $611.46CAD / Pkg of 1

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Versilon™ Silver Antimicrobial Tubing, 1/8" ID x 1/4" OD; 50 ft 5010714 Mfr # AS600007
COMPARE
1/8 1/4 1/16
$216.59CAD / Pkg of 1
Versilon™ Silver Antimicrobial Tubing, 3/16" ID x 5/16" OD; 50 ft 5010715 Mfr # AS600012
COMPARE
3/16 5/16 1/16
$255.26CAD / Pkg of 1
Versilon™ Silver Antimicrobial Tubing, 1/4" ID x 3/8" OD; 50 ft 5010716 Mfr # AS600017
COMPARE
1/4 3/8 1/16
$270.74CAD / Pkg of 1
Versilon™ Silver Antimicrobial Tubing, 3/8" ID x 1/2" OD; 50 ft 5010717 Mfr # AS600027
COMPARE
3/8 1/2 1/16
$379.03CAD / Pkg of 1
Versilon™ Silver Antimicrobial Tubing, 1/2" ID x 3/4 OD; 50 ft 5010718 Mfr # AS600038
COMPARE
1/2 3/4 1/8
$611.46CAD / Pkg of 1
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat