Versilon® FEP High-Purity Tubing
High-performance tubing exhibits excellent physical and chemical resistance
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • Made from chemically inert thermoplastic
  • Transparent and flexible—ideal for chemical transfer applications
  • Withstands temperatures up to 206°C (402°F)
  • Noncorroding, nonoxidizing, and UV resistant
The high-performance fluoropolymer tubing provides exceptional physical and chemical resistance, making it ideal for transferring most solvent, acids, and fuels. Made from chemically inert fluoropolymer, Versilon® FEP tubing will not corrode or oxidize. Flexible and transparent tubing is UV resistant and withstands temperatures up to 206°C (402°F).
$102.11 - $2,313.22CAD / Pkg of 1
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Versilon FEP Tubing, 1/16" ID x 1/8" OD; 50 Ft 5010719 Mfr # TSFE14-0125-031-50
COMPARE
1/16 1/8 1/32
$102.11CAD / Pkg of 1
Versilon FEP Tubing, 3/32" ID x 5/32" OD; 50 Ft 5010720 Mfr # TSFE14-0156-031-50
COMPARE
3/32 5/32 1/32
$142.33CAD / Pkg of 1
Versilon FEP Tubing, 1/16" ID x 3/16" OD; 50 Ft 5010721 Mfr # TSFE14-0187-062-50
COMPARE
1/16 3/16 1/16
$286.21CAD / Pkg of 1
Versilon FEP Tubing, 1/8" ID x 3/16" OD; 50 Ft 5010722 Mfr # TSFE14-0187-031-50
COMPARE
1/8 3/16 1/32
$177.91CAD / Pkg of 1
Versilon FEP Tubing, 3/16" ID x 1/4" OD; 50 Ft 5010723 Mfr # TSFE14-0250-031-50
COMPARE
3/16 1/4 1/32
$224.32CAD / Pkg of 1
Versilon FEP Tubing, 5/32" ID x 1/4" OD; 50 Ft 5010724 Mfr # TSFE14-0250-047-50
COMPARE
5/32 1/4 3/64
$317.15CAD / Pkg of 1
Versilon FEP Tubing, 1/8" ID x 1/4" OD; 50 Ft 5010725 Mfr # TSFE14-0250-062-50
COMPARE
1/8 1/4 1/16
$379.03CAD / Pkg of 1
Versilon FEP Tubing, 1/4" ID x 5/16" OD; 50 Ft 5010726 Mfr # TSFE14-0312-031-50
COMPARE
1/4 5/16 1/32
$286.21CAD / Pkg of 1
Versilon FEP Tubing, 3/16" ID x 5/16" OD; 50 Ft 5010727 Mfr # TSFE14-0312-062-50
COMPARE
3/16 5/16 1/16
$526.00CAD / Pkg of 1
Versilon FEP Tubing, 5/16" ID x 3/8" OD; 50 Ft 5010728 Mfr # TSFE14-0375-031-50
COMPARE
5/16 3/8 1/32
$355.82CAD / Pkg of 1
Versilon FEP Tubing, 1/4" ID x 3/8" OD; 50 Ft 5010729 Mfr # TSFE14-0375-062-50
COMPARE
1/4 3/8 1/16
$534.10CAD / Pkg of 1
Versilon FEP Tubing, 3/8" ID x 7/16" OD; 50 Ft 5010730 Mfr # TSFE14-0437-031-50
COMPARE
3/8 7/16 1/32
$409.97CAD / Pkg of 1
Versilon FEP Tubing, 7/16" ID x 1/2" OD; 50 Ft 5010731 Mfr # TSFE14-0500-031-50
COMPARE
7/16 1/2 1/32
$479.59CAD / Pkg of 1
Versilon FEP Tubing, 3/8" ID x 1/2" OD; 50 Ft 5010732 Mfr # TSFE14-0500-062-50
COMPARE
3/8 1/2 1/16
$758.80CAD / Pkg of 1
Versilon FEP Tubing, 1/2" ID x 9/16" OD; 50 Ft 5010733 Mfr # TSFE14-0562-031-50
COMPARE
1/2 9/16 1/32
$565.05CAD / Pkg of 1
Versilon FEP Tubing, 1/2" ID x 5/8" OD; 50 Ft 5010734 Mfr # TSFE14-0625-062-50
COMPARE
1/2 5/8 1/16
$957.70CAD / Pkg of 1
Versilon FEP Tubing, 5/8" ID x 3/4" OD; 50 Ft 5010735 Mfr # TSFE14-0750-062-50
COMPARE
5/8 3/4 1/16
$1,242.07CAD / Pkg of 1
Versilon FEP Tubing, 3/4" ID x 7/8" OD; 50 Ft 5010736 Mfr # TSFE14-0875-062-50
COMPARE
3/4 7/8 1/16
$1,768.07CAD / Pkg of 1
Versilon FEP Tubing, 7/8" ID x 1" OD; 50 Ft 5010737 Mfr # TSFE14-1000-062-50
COMPARE
7/8 1 1/16
$2,048.01CAD / Pkg of 1
Versilon FEP Tubing, 1" ID x 1-1/8" OD; 50 Ft 5010738 Mfr # TSFE14-1125-062-50
COMPARE
1 1 1/8 1/16
$2,313.22CAD / Pkg of 1
Showing 1- 20 of 20