Vanguard Safety Polyurethane Coated Cut Resistant Work Gloves
Palm coating resists abrasion, provides good wet and dry grip
  • Seamless knit HPPE blended liner keeps hands cool
  • FFDCA compliant for food handling
  • EN 388 Standard with 4X43C rating
  • ANSI abrasion level 5
  • ANSI A3 cut level
  • Machine washable
These A3 polyurethane palm coated work gloves are cut and abrasion resistant and offer excellent gripping while wet or dry. The breathable, seamless knit high performance polyethylene (HPPE) liner keeps your hands cool. The gloves are ideal for manufacturing, construction, metal and glass fabrication and machining work.
$985.70CAD / Case of 288
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Lining Size
Availability Pricing
Vanguard Safety R11A310 A3 HPPE PU Coated Cut Resistant Work Gloves, Salt & Pepper, XL; 144 Pairs/CS 7890711 Mfr # R11A310
COMPARE
9 27/32 13g High Performance Polyethylene (HPPE) Extra large
$985.70CAD / Case of 288
Vanguard Safety R11A36 A3 HPPE PU Coated Cut Resistant Work Gloves, Salt & Pepper, XS; 144 Pairs/CS 7890712 Mfr # R11A36
COMPARE
8 17/64 13g High Performance Polyethylene (HPPE) Extra small
$985.70CAD / Case of 288
Vanguard Safety R11A37 A3 HPPE PU Coated Cut Resistant Work Gloves, Salt & Pepper, Small; 144 Pairs/CS 7890713 Mfr # R11A37
COMPARE
7 7/8 13g High Performance Polyethylene (HPPE) Small
$985.70CAD / Case of 288
Vanguard Safety R11A38 A3 HPPE PU Coated Cut Resistant Work Gloves, Salt & Pepper, Medium; 144 Pairs/CS 7890714 Mfr # R11A38
COMPARE
9 13g High Performance Polyethylene (HPPE) Medium
$985.70CAD / Case of 288
Vanguard Safety R11A39 A3 HPPE PU Coated Cut Resistant Work Gloves, Salt & Pepper, Large; 144 Pairs/CS 7890715 Mfr # R11A39
COMPARE
9 29/64 13g High Performance Polyethylene (HPPE) Large
$985.70CAD / Case of 288