Vanguard Safety A3CH2 Powder-Free Latex Industrial Gloves, 14 mil, Blue
Extended cuff protects wrists and forearms
  • Texture fingertips for secure grip
  • Powder-free
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
  • Ambidextrous
These disposable latex industrial gloves feature a 11 ½” extended cuff for added protection of wrists and forearms. The 14-mil thick blue gloves are highly flexible for exceptional comfort and dexterity, powder-free to reduce skin irritation, and textured fingertips provide secure grip. Ideal for use in automotive, food service, food processing, industrial, janitorial, and manufacturing industries.
$244.58CAD / Case of 500
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Size
Availability Pricing
Vanguard Safety Powder-Free Latex Industrial Gloves, 14 mil, Blue, Medium; 500/CS RK-78906-37 Mfr # A3CH22
COMPARE
11 1/2 Latex Medium
$244.58CAD / Case of 500
Vanguard Safety Powder-Free Latex Industrial Gloves, 14 mil, Blue, Large; 500/CS RK-78906-38 Mfr # A3CH23
COMPARE
11 1/2 Latex Large
$244.58CAD / Case of 500
Vanguard Safety Powder-Free Latex Industrial Gloves, 14 mil, Blue, Extra-Large; 500/CS RK-78906-39 Mfr # A3CH24
COMPARE
11 1/2 Latex Extra Large
$244.58CAD / Case of 500