Vanguard Safety A16 Nitrile Exam Gloves

Maintain dexterity without sacrificing safety
  • Powder-free—made of soft 100% nitrile
  • Latex-free to eliminate adverse reactions
  • Textured surface for better gripping
  • Meets acceptable quality level (AQL)
  • Chemotherapy rated
  • FDA approved
  • Medical grade
  • Ambidextrous
These Vanguard Safety A16 nitrile exam gloves are made from powder-free and latex-free nitrile. They feature textured surface for improved grip and maximum ease of use with delicate instruments. All A16 nitrile gloves are ambidextrous and blue in color. Gloves are packed 100/box and 10 boxes per case.
$138.50CAD / Case of 1000

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Size Thickness (mil) Description
Availability Pricing
Vanguard Safety A16 Nitrile Exam Gloves, Small; 1000/CS RK-78902-05 Mfr # A16A11
COMPARE
Small 5 Nitrile Exam Gloves, Small; 1000/Cs
$138.50CAD / Case of 1000
Vanguard Safety A16 Nitrile Exam Gloves, Medium; 1000/CS RK-78902-06 Mfr # A16A12
COMPARE
Medium 5 Nitrile Exam Gloves, Medium, 1000/Cs
$138.50CAD / Case of 1000
Vanguard Safety A16 Nitrile Exam Gloves, Large; 1000/CS RK-78902-07 Mfr # A16A13
COMPARE
Large 5 Nitrile Exam Gloves, Large; 1000/Cs
$138.50CAD / Case of 1000
Vanguard Safety A16 Nitrile Exam Gloves, X-Large; 1000/CS RK-78902-08 Mfr # A16A14
COMPARE
X-Large 5 Nitrile Exam Gloves, X-Large; 1000/Cs
$138.50CAD / Case of 1000
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat