Unico UV/Visible Spectrophotometers
Affordable performance – with optional multicell holder
  • Store up to 200 test methods for your demanding lab environments
  • Software-driven DNA/RNA/protein/kinetics analysis
The 4 nm bandwidth of these single-beam instruments produces precise response to transmittance changes for UV/visible spectrum methods. Four-position cuvette holders save time and improve efficiency.
Use the RS-232 or USB ports for connecting directly to a printer or to a PC. Order optional Peltier temperature controller and/or sipper system for total spectrophotometric utility. Peltier controller regulates temperature between 15 and 40°C.
UV/Visible spectrophotometers have a low profile with a large LED display which can be read from any angle. Optional software 83057-65 lets you create standard curves and absorbance vs. time graphs and also includes a wavelength scanning feature.
$9,204.27CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Wavelength (nm) Power (VAC)
Availability Pricing
Unico UV/Visible Spectrophotometer, 230 VAC, 50 Hz 8305915 Mfr # CP-IS215UE
COMPARE
1000 230
$9,204.27CAD / Each