Cannon® Ubbelohde Calibrated Glass Capillary Viscometers
Determine viscosities using ASTM testing methods
  • Each viscometer is calibrated in accordance with ASTM D446 Comes with a calibration certificate that includes the viscometer constant and formulas for calculating viscosities
Use to determine kinematic viscosities of transparent Newtonian liquids. Require an 11-mL sample volume. Precision is ±0.2%.
$400.76 - $415.50CAD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Approximate Constant (cSt/sec) Max Viscosity Range (cSt) Universal Size Number
Availability Pricing
Cannon 9721-R50 Viscometer, Size 0; 0.3 to 1 Centistokes RK-98934-10 Mfr # 9721-R50
COMPARE
0.001 1 0
$400.76CAD / Each
Cannon 9721-R53 Viscometer, Size 0C; 0.6 to 3 Centistokes RK-98934-11 Mfr # 9721-R53
COMPARE
0.003 3 0C
$411.08CAD / Each
Cannon 9721-R56 Viscometer, Size 0B; 1 to 5 Centistokes RK-98934-12 Mfr # 9721-R56
COMPARE
0.005 5 0B
$411.08CAD / Each
Cannon 9721-R59 Viscometer, Size 1; 2 to 10 Centistokes RK-98934-13 Mfr # 9721-R59
COMPARE
0.01 10 1
$411.08CAD / Each
Cannon 9721-R62 Viscometer, Size 1C; 6 to 30 Centistokes RK-98934-14 Mfr # 9721-R62
COMPARE
0.03 30 1C
$411.08CAD / Each
Cannon 9721-R65 Viscometer, Size 1B; 10 to 50 Centistokes RK-98934-15 Mfr # 9721-R65
COMPARE
0.05 50 1B
$411.08CAD / Each
Cannon 9721-R68 Viscometer, Size 2; 20 to 100 Centistokes RK-98934-16 Mfr # 9721-R68
COMPARE
0.1 100 2
$411.08CAD / Each
Cannon 9721-R71 Viscometer, Size 2C; 60 to 300 Centistokes RK-98934-17 Mfr # 9721-R71
COMPARE
0.3 300 2C
$415.50CAD / Each
Cannon 9721-R74 Viscometer, Size 2B; 100 to 500 Centistokes RK-98934-18 Mfr # 9721-R74
COMPARE
0.5 500 2B
$411.08CAD / Each
Cannon 9721-R77 Viscometer, Size 3; 200 to 1000 Centistokes RK-98934-19 Mfr # 9721-R77
COMPARE
1 1000 3
$415.50CAD / Each
Cannon 9721-R80 Viscometer, Size 3C; 600 to 3000 Centistokes RK-98934-20 Mfr # 9721-R80
COMPARE
3 3000 3C
$415.50CAD / Each
Cannon 9721-R83 Viscometer, Size 3B; 1000 to 5000 Centistokes RK-98934-21 Mfr # 9721-R83
COMPARE
5 5000 3B
$400.76CAD / Each
Cannon 9721-R86 Viscometer, Size 4; 2000 to 10,000 Centistokes RK-98934-22 Mfr # 9721-R86
COMPARE
10 10000 4
$400.76CAD / Each
Cannon 9721-R89 Viscometer, Size 4C; 6000 to 30,000 Centistokes RK-98934-23 Mfr # 9721-R89
COMPARE
30 30000 4C
$405.18CAD / Each
Cannon 9721-R92 Viscometer, Size 4B; 10,000 to 50,000 Centistokes RK-98934-24 Mfr # 9721-R92
COMPARE
50 50000 4B
$400.76CAD / Each
Cannon 9721-R95 Viscometer, Size 5; 20,000 to 100,000 Centistokes RK-98934-25 Mfr # 9721-R95
COMPARE
100 100000 5
$403.71CAD / Each
Showing 1- 16 of 16