Tygon® E-3603 Laboratory Tubing
Nonoxidizing and noncontaminating—ideal for general laboratory applications
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • Smooth, polished inner wall prevents buildups
  • Excellent lot-to-lot consistency for reproducible results
  • Great flex fatigue resistance
  • BPA free
Use the clear and flexible Tygon® E-3603 tubing for various general laboratory applications. This nonoxidizing and noncontaminating tubing offers excellent chemical resistance to virtually all organic chemicals used in the laboratory. High tolerances and lot-to-lot consistency ensure reproducible results. Tubing slips over fittings and grips tight for a secure fit, making laboratory setup quick and simple. Abrasion and flex-fatigue resistant tubing provides superior life span.
$19.30 - $2,114.31CAD / Pkg of 1
32 variations of this product are available.
See All Product Options Below
32 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon E-3603 Tubing, 1/32" ID x 3/32" OD; 50 Ft 5010620 Mfr # ACF00001
COMPARE
1/32 3/32 1/32
$19.30CAD / Pkg of 1
Tygon E-3603 Tubing, 1/16" ID x 1/8" OD; 50 Ft 5010621 Mfr # ACF00002
COMPARE
1/16 1/8 1/32
$31.24CAD / Pkg of 1
Tygon E-3603 Tubing, 1/16" ID x 3/16" OD; 50 Ft 5010622 Mfr # ACF00003
COMPARE
1/16 3/16 1/16
$78.09CAD / Pkg of 1
Tygon E-3603 Tubing, 3/32" ID x 5/32" OD; 50 Ft 5010623 Mfr # ACF00004
COMPARE
3/32 5/32 1/32
$40.67CAD / Pkg of 1
Tygon E-3603 Tubing, 3/32" ID x 7/32" OD; 50 Ft 5010624 Mfr # ACF00005
COMPARE
3/32 7/32 1/16
$99.01CAD / Pkg of 1
Tygon E-3603 Tubing, 1/8" ID x 3/16" OD; 50 Ft 5010625 Mfr # ACF00006
COMPARE
1/8 3/16 1/32
$48.47CAD / Pkg of 1
Tygon E-3603 Tubing, 1/8" ID x 1/4" OD; 50 Ft 5010626 Mfr # ACF00007
COMPARE
1/8 1/4 1/16
$116.10CAD / Pkg of 1
Tygon E-3603 Tubing, 5/32" ID x 7/32" OD; 50 Ft 5010627 Mfr # ACF00009
COMPARE
5/32 7/32 1/32
$61.00CAD / Pkg of 1
Tygon E-3603 Tubing, 3/16" ID x 5/16" OD; 50 Ft 5010628 Mfr # ACF00012
COMPARE
3/16 5/16 1/16
$162.44CAD / Pkg of 1
Tygon E-3603 Tubing, 3/16" ID x 3/8" OD; 50 Ft 5010629 Mfr # ACF00013
COMPARE
3/16 3/8 3/32
$263.00CAD / Pkg of 1
Tygon E-3603 Tubing, 1/4" ID x 3/8" OD; 50 Ft 5010630 Mfr # ACF00017
COMPARE
1/4 3/8 1/16
$201.12CAD / Pkg of 1
Tygon E-3603 Tubing, 1/4" ID x 7/16" OD; 50 Ft 5010631 Mfr # ACF00018
COMPARE
1/4 7/16 3/32
$317.15CAD / Pkg of 1
Tygon E-3603 Tubing, 1/4" ID x 1/2" OD; 50 Ft 5010632 Mfr # ACF00019
COMPARE
1/4 1/2 1/8
$379.03CAD / Pkg of 1
Tygon E-3603 Tubing, 5/16" ID x 7/16" OD; 50 Ft 5010633 Mfr # ACF00022
COMPARE
5/16 7/16 1/16
$232.06CAD / Pkg of 1
Tygon E-3603 Tubing, 5/16" ID x 1/2" OD; 50 Ft 5010634 Mfr # ACF00023
COMPARE
5/16 1/2 3/32
$332.62CAD / Pkg of 1
Tygon E-3603 Tubing, 5/16" ID x 9/16" OD; 50 Ft 5010635 Mfr # ACF00024
COMPARE
5/16 9/16 1/8
$394.50CAD / Pkg of 1
Tygon E-3603 Tubing, 3/8" ID x 1/2" OD; 50 Ft 5010636 Mfr # ACF00027
COMPARE
3/8 1/2 1/16
$263.00CAD / Pkg of 1
Tygon E-3603 Tubing, 3/8" ID x 9/16" OD; 50 Ft 5010637 Mfr # ACF00028
COMPARE
3/8 9/16 3/32
$340.35CAD / Pkg of 1
Tygon E-3603 Tubing, 3/8" ID x 5/8" OD; 50 Ft 5010638 Mfr # ACF00029
COMPARE
3/8 5/8 1/8
$495.06CAD / Pkg of 1
Tygon E-3603 Tubing, 7/16" ID x 9/16" OD; 50 Ft 5010639 Mfr # ACF00032
COMPARE
7/16 9/16 1/16
$309.41CAD / Pkg of 1
Showing 1 - 20 of 32