Tygon® E-1000 Food and Beverage Dispensing Tubing
Extremely low durometer—ideal for use in low-torque pumps
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • Remains flexible even at -55°C (-67°F)
  • Soft and flexible tubing resists twisting and collapsing
  • Non-DEHP tubing formulation
  • Resistant to corrosive chemicals
Specially formulated for food and beverage dispensers, Tygon® E-1000 tubing offers high performance even at temperatures as low as -55°C (-67°F). Soft and super flexible, tubing easily manages sharp radius curves and multiple directional changes without twisting and collapsing. The low-durometer tubing exhibits minimal resistance to compression, making it ideal for low-torque or battery-operated peristaltic pumps. Made with non-DEHP [bis (2-ethylhexyl) phthalate] plasticizer, Tygon E-1000 tubing exhibits excellent resistance to corrosive chemicals.
$95.77 - $744.06CAD / Pkg of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon E-1000 Tubing, 1/16" ID x 3/16" OD; 50 Ft 5010610 Mfr # ADK00003
COMPARE
1/16 3/16 1/16
$95.77CAD / Pkg of 1
Tygon E-1000 Tubing, 1/8" ID x 1/4" OD; 50 Ft 5010611 Mfr # ADK00007
COMPARE
1/8 1/4 1/16
$139.97CAD / Pkg of 1
Tygon E-1000 Tubing, 3/16" ID x 5/16" OD; 50 Ft 5010612 Mfr # ADK00012
COMPARE
3/16 5/16 1/16
$198.91CAD / Pkg of 1
Tygon E-1000 Tubing, 1/4" ID x 3/8" OD; 50 Ft 5010613 Mfr # ADK00017
COMPARE
1/4 3/8 1/16
$243.11CAD / Pkg of 1
Tygon E-1000 Tubing, 1/4" ID x 1/2" OD; 50 Ft 5010614 Mfr # ADK00019
COMPARE
1/4 1/2 1/8
$471.48CAD / Pkg of 1
Tygon E-1000 Tubing, 5/16" ID x 7/16" OD; 50 Ft 5010615 Mfr # ADK00022
COMPARE
5/16 7/16 1/16
$279.94CAD / Pkg of 1
Tygon E-1000 Tubing, 3/8" ID x 1/2" OD; 50 Ft 5010616 Mfr # ADK00027
COMPARE
3/8 1/2 1/16
$324.15CAD / Pkg of 1
Tygon E-1000 Tubing, 3/8" ID x 5/8" OD; 50 Ft 5010617 Mfr # ADK00029
COMPARE
3/8 5/8 1/8
$604.09CAD / Pkg of 1
Tygon E-1000 Tubing, 1/2" ID x 5/8" OD; 50 Ft 5010618 Mfr # ADK00036
COMPARE
1/2 5/8 1/16
$397.82CAD / Pkg of 1
Tygon E-1000 Tubing, 1/2" ID x 3/4" OD; 50 Ft 5010619 Mfr # ADK00038
COMPARE
1/2 3/4 1/8
$744.06CAD / Pkg of 1
Showing 1- 10 of 10