Troemner UltraClass Analytical Calibration Masses
UltraClass - The weight tolerance is equal to ANSI/ASTM E617 Class 0 and exceeds OIML R 111 Class E2. This class is used as a reference standard for calibrating other reference standards and is appropriate for calibrating highprecision analytical balances with a readability as low as 0.01 mg.
  • UltraClass Tolerance = ASTM Class 0
  • UltraClass weights, 1 g through 5 kg, are made from exclusive Troemner Alloy 8 stainless steel
These weights are developed to meet the most demanding calibration needs with the ability to be adjusted. Consistent automated measurement means that Troemner UltraClass weights are the most precise two-piece weights available with weight tolerances that equal or exceed comparable ANSI/ASTM E617 Classes. The weights are manufactured of the finest stainless steels available, including Troemner Alloy 8 Stainless Steel, which is available exclusively through Troemner. This unique material possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion. Alloy 8 is the finest grade of stainless steel available and is similar to material originally specified in the manufacture of 1 kg state lab primary standards.Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerancesdecrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.Suggested Markets for Analytical Precision Weights include, but are not limited to, pharmaceutical, biotech, nanotechnology, life science, forensic and nuclear industries.
$243.85 - $2,058.33CAD / Each
29 variations of this product are available.
See All Product Options Below
29 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (g)
Availability Pricing
Troemner 7037-0W 1 mg, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102097 Mfr # 7037-0W
COMPARE
NVLAP 0.001
$286.94CAD / Each
Troemner 7036-0W 2 mg, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102098 Mfr # 7036-0W
COMPARE
NVLAP 0.002
$286.94CAD / Each
Troemner 7033-0W 10 mg, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102101 Mfr # 7033-0W
COMPARE
NVLAP 0.01
$286.94CAD / Each
Troemner 7032-0W 20 mg, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102102 Mfr # 7032-0W
COMPARE
NVLAP 0.02
$286.94CAD / Each
Troemner 7029-0W 100 mg, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102104 Mfr # 7029-0W
COMPARE
NVLAP 0.1
$286.94CAD / Each
Troemner 7028-0W 200 mg, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102105 Mfr # 7028-0W
COMPARE
NVLAP 0.2
$286.94CAD / Each
Troemner 7021-0W 10 g, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102711 Mfr # 7021-0W
COMPARE
NVLAP 10
$318.25CAD / Each
Troemner 7022-0W 5 g, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102713 Mfr # 7022-0W
COMPARE
NVLAP 5
$318.25CAD / Each
Troemner 7025-0W 1 g, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102717 Mfr # 7025-0W
COMPARE
NVLAP 1
$318.25CAD / Each
Troemner 7020-0W 20 g, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102737 Mfr # 7020-0W
COMPARE
NVLAP 20
$337.41CAD / Each
Troemner 7018-0W 50 g, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102739 Mfr # 7018-0W
COMPARE
NVLAP 50
$365.03CAD / Each
Troemner 7017-0W 100 g, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102741 Mfr # 7017-0W
COMPARE
NVLAP 100
$404.08CAD / Each
Troemner 7016-0W 200 g, UltraClass Analytical Mass with NVLAP Accredited Certificate 0102743 Mfr # 7016-0W
COMPARE
NVLAP 200
$621.40CAD / Each
Troemner 7012-0T 2 kg, UltraClass Analytical Mass with Traceable Certificate 0103102 Mfr # 7012-0T
COMPARE
NIST 2000
$2,058.33CAD / Each
Troemner 7014-0T 500 g, UltraClass Analytical Mass with Traceable Certificate 0103104 Mfr # 7014-0T
COMPARE
NIST 500
$675.18CAD / Each
Troemner 7016-0T 200 g, UltraClass Analytical Mass with Traceable Certificate 0103106 Mfr # 7016-0T
COMPARE
NIST 200
$495.06CAD / Each
Troemner 7017-0T 100 g, UltraClass Analytical Mass with Traceable Certificate 0103107 Mfr # 7017-0T
COMPARE
NIST 100
$338.88CAD / Each
Troemner 7020-0T 20 g, UltraClass Analytical Mass with Traceable Certificate 0103109 Mfr # 7020-0T
COMPARE
NIST 20
$305.73CAD / Each
Troemner 7021-0T 10 g, UltraClass Analytical Mass with Traceable Certificate 0103111 Mfr # 7021-0T
COMPARE
NIST 10
$305.73CAD / Each
Troemner 7022-0T 5 g, UltraClass Analytical Mass with Traceable Certificate 0103112 Mfr # 7022-0T
COMPARE
NIST 5
$305.73CAD / Each
Showing 1 - 20 of 29