Troemner Class-F Avoirdupois Stainless Steel Cylindrical Test Weights
Stainless Steel Test Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians.
Stainless Steel Cylindrical Weights are legal for trade, which means their design meets specifications as outlined in NIST Handbook 105-1 and are approved to be used on legal for trade devices within the United States. Weights 5 kg/10 lb and below must be made of stainless steel of quality no lower than 300 series. Adjusting cavities are typically on the top of the weight and must be tamper resistant.
Stainless Steel Class F Test Weights may be used to test most accuracy Class III scales, all scales of Class IIIL or IIII, and scales not marked with a class designation. Class F is also acceptable
for use in verifying scales that have 0.1% accuracy or lower. Class F Test Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians. Suggested markets for Stainless Steel Test Weights include, but are not limited to, Hospitals and Food Services.
$130.54 - $574.62CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (lbs) Description
Availability Pricing
Troemner Test Weight 1 Lb Class F with Traceable Certificate, Cyl Indrical with Groove 0102377 Mfr # 1211T
COMPARE
NIST 1 1 Lb Class F with Traceable Certificate, Cyl Indrical with Groove
$323.41CAD / Each
Troemner Test Weight 2 Lb Class F with Traceable Certificate, Cyl Indrical with Groove 0102378 Mfr # 1209T
COMPARE
NIST 2 2 Lb Class F with Traceable Certificate, Cyl Indrical with Groove
$352.51CAD / Each
Troemner Test Weight 3 Lb Class F with Traceable Certificate, Cyl Indrical with Groove 0102379 Mfr # 1207T
COMPARE
NIST 3 3 Lb Class F with Traceable Certificate, Cyl Indrical with Groove
$422.49CAD / Each
Troemner Test Weight 4 Lb Class F with Traceable Certificate, Cyl Indrical with Groove 0102380 Mfr # 1205T
COMPARE
NIST 4 4 Lb Class F with Traceable Certificate, Cyl Indrical with Groove
$436.86CAD / Each
Troemner Test Weight 5 Lb Class F with Traceable Certificate, Cyl Indrical with Groove 0102381 Mfr # 1203T
COMPARE
NIST 5 5 Lb Class F with Traceable Certificate, Cyl Indrical with Groove
$450.86CAD / Each
Troemner Test Weight 10lb Class F with Traceable Certificate 0102382 Mfr # 1201T
COMPARE
NIST 10 10lb Class F with Traceable Certificate
$574.62CAD / Each
Troemner 1201 10 lb Class F Stainless Steel Test Weight with No Certificate, Cylindrical with Groove 0103394 Mfr # 1201
COMPARE
None 10 10 lb Class F Stainless Steel Test Weight with No Certificate, Cylindrical with Groove
$446.81CAD / Each
Troemner 1203 5 lb Class 6 Stainless Steel Test Weight with No Certificate, Cylindrical with Groove 0103395 Mfr # 1203
COMPARE
None 5 5 lb Class 6 Stainless Steel Test Weight with No Certificate, Cylindrical with Groove
$289.15CAD / Each
Troemner 1205 4 lb Class F Stainless Steel Test Weight with No Certificate, Cylindrical with Groove 0103396 Mfr # 1205
COMPARE
None 4 4 lb Class F Stainless Steel Test Weight with No Certificate, Cylindrical with Groove
$273.31CAD / Each
Troemner 1207 3 lb Class F Stainless Steel Test Weight with No Certificate, Cylindrical with Groove 0103397 Mfr # 1207
COMPARE
None 3 3 lb Class F Stainless Steel Test Weight with No Certificate, Cylindrical with Groove
$254.90CAD / Each
Troemner 1209 2 lb Class F Stainless Steel Test Weight with No Certificate, Cylindrical with Groove 0103398 Mfr # 1209
COMPARE
None 2 2 lb Class F Stainless Steel Test Weight with No Certificate, Cylindrical with Groove
$180.49CAD / Each
Troemner 1211 1 lb Class F Stainless Steel Test Weight with No Certificate, Cylindrical with Groove 0103399 Mfr # 1211
COMPARE
None 1 1 lb Class F Stainless Steel Test Weight with No Certificate, Cylindrical with Groove
$130.54CAD / Each
Showing 1- 12 of 12