Trajan Splitless Inlet Liner with Double Taper

Spot the Difference in the SGE Lineup!
  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
Bottom taper minimizes contact with metal parts of the inlet and focuses sample onto the column head. Top taper aids in minimizing sample flashback
$276.63 - $1,088.84CAD / Pkg of 5

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 092018 Split/Splitless liner with double taper, Compatible with Agilent Systems, 78.5 mm L; 5/Pk 1600942 Mfr # 092018
COMPARE
$276.63CAD / Pkg of 5
Trajan SGE 092230 Split/Splitless Liner with double taper, Compatible with Agilent Systems, 78.5 mm L; 25/Pk 1601079 Mfr # 092230
COMPARE
$1,088.84CAD / Pkg of 25
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat