Traceable® Ultra™ Refrigerator/Freezer Thermometers with Calibration
Smart-Alarm™ signals both audible and visual alarms even after return to normal conditions
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Triple display simultaneously shows MIN/MAX and current temperature
  • 10-ft microcable allows refrigerator/freezer doors to close on it
Accurately monitor temperatures in refrigerators, freezers, water baths, and incubators. Thermometers have accuracy of ±0.5°C (-30 to 50°C). Built-in audible and visual alarms activate when temperature rises or falls below the alarm set point, and is programmable in 0.1° increments.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$103.14 - $134.08CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number Of Probe Channels Types Of Probes Accepted Description
Availability Pricing
COMPARE
1 1 bottle probe Calibrated Refrigerator/Freezer Thermometer; 1 Bottle Probe
$110.50CAD / Each
COMPARE
2 2 bottle probes Calibrated Refrigerator/Freezer Thermometer; 2 Bottle Probes
$128.92CAD / Each
COMPARE
1 1 Bullet probe Calibrated Refrigerator/Freezer Thermometer; 1 Bullet Probe
$103.14CAD / Each
COMPARE
2 2 Bullet probes Calibrated Refrigerator/Freezer Thermometer; 2 Bullet Probes
$106.08CAD / Each
COMPARE
1 1 bottle and 1 Bullet probes Calibrated Refrigerator/Freezer Thermometer; 1 Bottle and 1 Bullet Probe
$111.98CAD / Each
COMPARE
1 1 vaccine bottle probe Calibrated Refrigerator/Freezer Thermometer; 1 Vaccine Bottle Probe
$109.77CAD / Each
COMPARE
2 2 vaccine bottle probes Calibrated Refrigerator/Freezer Thermometer; 2 Vaccine Bottle Probes
$129.66CAD / Each
COMPARE
1 1 plastic bottle probe Calibrated Refrigerator/Freezer Thermometer; 1 Plastic Bottle Probe
$113.45CAD / Each
COMPARE
2 2 plastic bottle probes Calibrated Refrigerator/Freezer Thermometer; 2 Plastic Bottle Probes
$134.08CAD / Each
Showing 1- 9 of 9