Traceable® Time and Date Digital Thermometers with Calibration
Minimum and maximum readings are time/date stamped
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Simultaneously displays temperatures and time
  • Wire mounting bracket permits easy probe placement
Ideal for monitoring temperature in the lab, home or office. Thermometer includes a remote probe with 6-ft (1.8m) cable to measure outdoor temperature and an internal sensor to measure indoor temperature. Display can be changed to show probe or ambient temperature, time of day, and month/day.
Thermometer can also display the exact time and date when the minimum and maximum temperature occurred. The temperature sensor performs accurately in most environments even including under water. Use rear stand to place unit on table or attach to wall via hook-and-loop strip or the two rear-mounted screw holes.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$72.20 - $85.46CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Description
Availability Pricing
COMPARE
176 Calibrated Time and Date Digital Thermometer; 1 Wire Probe
$72.20CAD / Each
COMPARE
176 Calibrated Time and Date Digital Thermometer; 1 Bottle Probe
$85.46CAD / Each
COMPARE
176 Calibrated Time and Date Digital Thermometer; 1 5-mL Bottle Probe
$81.04CAD / Each