Torbal FSC Tool & Wrench Torque Tester
Six built in modes and applications
  • 1000Hz data capture
  • 2.75" LCD display with backlight
  • Wireless, USB, RS232 and analog l/O interfaces
  • Onscreen stats and analytics
  • Onscreen graphing and histogram
These advanced precision torque testers provide accurate verification or adjustment of virtualy any manual or electric wrench, and screwdriver. Heavy duty design makes thes FSC torque gauge ideal for any fastener assembly application. The realiable design, ease of use, and adaptability to existing workbenches or tabletops improves productivity. The loadcell socket accepts any 1/4" drive wrench and 1/4" to 3/8" drive adapters. Easily connect and communicate with PC or peripherals with wireless, USB, RS232, and analog l/O interfaces. Ave and transfer data to a PC with the removable MocroSD memory card.
$2,305.86 - $2,388.37CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-in) Capacity (N-m) Grads.
Availability Pricing
Torbal FSC5 Advanced Precision Tool & Wrench Torque Tester, 44 lb in capacity 2530480 Mfr # FSC5
COMPARE
44 5 Nm, N.cm, kgf.m, gf.m, lbf.in
$2,305.86CAD / Each
Torbal FSC10 Advanced Precision Tool & Wrench Torque Tester, 100 lb in capacity 2530481 Mfr # FSC10
COMPARE
100 10 Nm, N.cm, kgf.m, gf.m, lbf.in
$2,388.37CAD / Each