Tintometer PFXi-195 Series Short-Pathlength Color Meters
Eliminate user subjectivity with unbiased color measurements based on reference standards
  • Large graphic LCD with user-friendly prompts in several languages
  • Extensive choice of standard color scales or customized color scales from reference samples
  • Easily removable sample chamber offering a range of sample cell sizes
  • Data output meets GLP standards with date, time, sample, and user ID
Spectrophotometric color meters provide accurate, reliable color data from optically clear samples. The meter automatically measures color transmittance and displays results directly in industry color scales, CIE values, and as spectral data. The direct measurements on large LCD supply unbaised readings, unaffected by operator or environment.
The short-pathlength meter is ideal for slightly colored solutions, like wastewater, syrups, honey, dark oils, and beers. The meter offers built-in menu guides to help navigate setup parameters for the different color scales. The meter performance is verified with self-diagnostic check tests, and accuracy is ensured with calibrations using certified glass filter(s) included with meter (filters differ depending upon model).
Flexible data transfer with USB, LAN, and RS-232 interfaces. Remote calibration and maintenance service are also available via the internet. Contact our Applications Specialists for upgrades to color scales.
$22,802.11 - $24,127.94CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Lovibond PFXi-195/1 Short Pathlength Color Meter, Liquid Chemicals and Oils 9956202 Mfr # 1371951
COMPARE
Short Pathlength Color Meter, Liquid Chemicals and Oils
$22,802.11CAD / Each
Lovibond PFXi-195/2 Petroleum, Oils, and Fuels Short Pathlength Color Meter 9956204 Mfr # 1371952
COMPARE
Petroleum, Oils, and Fuels Short Pathlength Color Meter
$24,127.94CAD / Each