ThermoProbe Heavy-Duty Tank Gauging Thermometers

Intrinsically safe thermometer for accurate temperature measurement in bulk liquid storage tanks
  • Ideal alternative to hazardous glass thermometers
  • Reliable and stable RTD sensor technology
  • Accuracy ±0.2°F from 32 to 200°F (0 to 100°C)
  • Easy to calibrate and make adjustments using faceplate controls
  • Cable marked every 5 feet
  • Includes 4-point NIST traceable calibration report
The ThermoProbe are designed as rugged, field-use, portable precision digital thermometers, involving volume-temperature gauging of petroleum and chemical products. Convenient design allows for easy retrieval of the cable probe and eliminates the need to bring the probe to the surface to take multiple readings. Typical applications include custody transfers, tanks, pipelines, ships, railcars, barges, tanker trucks, and proving systems.

Zone 0 intrinsically safe meters are manufactured of materials which are both immune to petrochemicals and are nonsparking. The probe assembly is constructed with a very flexible, static-dissipating, aramid fiber reinforced, nonstick cable and stainless steel components.

Large LCD displays temperature, plus arrows indicate the direction of the temperature reading and whether stability has been reached. Stabilized temperatures can be logged at numerous liquid levels for a running average and later displayed for documenting. Advanced features, accessible through a user-friendly menu, include Min/Max/Avg reading, readout integrity, photosensor-controlled backlight, selectable (°F/°C) unit of measure, battery status, and automatic shut-off.
$2,418.42 - $3,077.91CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Description
Availability Pricing
ThermoProbe TP7D-050-SW-SM Spool-Type 50ft Std Probe 9002601 Mfr # TP7D-050-SW-SM
COMPARE
400 RTD Spool-Type 50ft Std Probe
$2,858.08CAD / Each
ThermoProbe TP7D-075-SW-SM Spool-Type 75ft Std Probe 9002602 Mfr # TP7D-075-SW-SM
COMPARE
400 RTD Spool-Type 75ft Std Probe
$2,572.24CAD / Each
ThermoProbe TP7D-110-EX-SM Spool-Type 110ft Extra Weighted Probe 9002603 Mfr # TP7D-110-EX-SM
COMPARE
400 RTD Spool-Type 110ft Extra Weighted Probe
$3,077.91CAD / Each
ThermoProbe TP9A-050-SW-SM Stainless Steel 50ft Std Probe 9002604 Mfr # TP9A-050-SW-SM
COMPARE
400 RTD Stainless Steel 50ft Std Probe
$2,968.07CAD / Each
ThermoProbe TP9-075A-SW-SM Stainless Steel 75ft Std Probe 9002605 Mfr # TP9A-075-SW-SM
COMPARE
400 RTD Stainless Steel 75ft Std Probe
$2,418.42CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat