Thermo Scientific SAHARA Premium Stainless Steel Heating Circulating Baths
Superior performance for the most demanding applications
  • Ramp programming (PC 200, 5x10 segments; PC 300, 10x10 segments), with edit during ramp feature
  • Large color LCD (262K colors) — graphical display of operating ramp program
  • Pump speed adjustment to 10 levels for turbulence control
Building on the capabilities of the Standard (SC) and Advanced (AC) controllers in the SAHARA family, the Premium (PC) model encompasses all of the features of the SC and AC baths, but then provides greater control and flexibility. Use the 10-key access keypad for fast and intuitive software navigation. Application identification (Unit control parameters are optimized for the application behavior, so that temperature control can be as fast as possible with zero over swing or under swing). User level management, e.g. for application temperature limitation: set high and low temperature limits to the system not allowing another user to set the temperature higher or lower than is established. Data logging: all ring buffer data will be stored for serial communication. Extra communication option for Profibus, CAN bus Analog I/O. User selectable high temperature cut-off (only PC 300).
$8,703.31 - $10,033.79CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
Thermo Scientific PC200-S7 8L Heated Bath, Amb +13 to 200C, 115V 1214800 Mfr # 1571072
COMPARE
200 115 8
$8,703.31CAD / Each
Thermo Scientific PC 300-S13 12L Heated Bath, Amb +13 to 300C, 208V 1214826 Mfr # 1591135
COMPARE
300 230 12
$10,033.79CAD / Each