Thermo Scientific Orion Half-Cell Ion-Selective Electrodes
For a complete system:
  • Ion-selective electrode
  • Reference electrode
  • Standardizing solution
  • Filling solution for reference electrodes
  • Ionic strength and pH adjustors
Select the ion-selective electrodes, reference electrodes, and necessary solutions to best fit your application. Choose ion-selective electrodes with BNC connectors and 3-ft cord. Note: All electrodes listed are sensing half-cells which require a reference electrode.
$1,599.66 - $1,919.90CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Ion-Selective Electrode
Availability Pricing
Thermo Scientific 9409BN Solid State Half-Cell ISE Electrode; Fluoride 0572111 Mfr # 9409BN
COMPARE
Fluoride
$1,919.90CAD / Each
Thermo Scientific 9417BN Solid State Half-Cell ISE Electrode; Chloride 0572141 Mfr # 9417BN
COMPARE
Chloride
$1,599.66CAD / Each