Thermo Scientific™ Nalgene™ Rectangular Carboys, HDPE
Don’t waste valuable bench space
  • Space-saving design for no dead area like traditional round carboys
  • Both liter and gallon graduations for easy volume determination
  • Leakproof high-density polyethylene (HDPE) for safe transport
Space-saving leakproof rectangular carboys feature liter/gallon graduations and built-in shoulder loops with stainless steel handles. Carboys with quick-action, needle-type spigots accept 5/8" ID tubing and come with adapters for ¼" or 5/16" ID tubing, integrally molded threaded boss, and two PTFE O-rings for additional versatility. Aspirator carboys feature integral tubulation for ½" ID tubing.
$265.21 - $475.90CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene 2211-0020 Rectangular HDPE Carboy with Handle, 9 L RK-06032-20 Mfr # 2211-0020
COMPARE
9 100 mm High Density Polyethyene (HDPE)
$265.21CAD / Each
Thermo Scientific Nalgene 2211-0050 Rectangular HDPE Carboy; 20 L RK-06032-50 Mfr # 2211-0050
COMPARE
20 100 mm High Density Polyethyene (HDPE)
$368.35CAD / Each
Thermo Scientific Nalgene 2320-0020 Rectangular HDPE Carboy with Spigot, 9 L RK-06066-20 Mfr # 2320-0020
COMPARE
9 100 mm High Density Polyethyene (HDPE)
$380.13CAD / Each
Thermo Scientific Nalgene 2320-0050 Rectangular HDPE Carboy with Spigot, 20 L RK-06066-50 Mfr # 2320-0050
COMPARE
20 100 mm High Density Polyethyene (HDPE)
$475.90CAD / Each