Thermo Scientific Nalgene Calibrated Graduated Polymethylpentene (PMP) Cylinders
Transparent polymethylpentene (PMP) cylinders offer good chemical and heat resistance
  • Meet ASTM Class B, E1272 specifications and ISO Standard 6706
Transparent polymethylpentene (PMP) cylinders have molded graduations. Resist dyes and stains. Calibrated TC/TD at 20°>C. Nonwetting; no meniscus. Large, stable PP base. Autoclavable, but may affect accuracy. Call for additional sizes.
$61.59 - $626.19CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Thermo Scientific Nalgene 3663-0010 Clear PMP Graduated Cylinder, 10 mL, 1/Pk 0613410 Mfr # 3663-0010
COMPARE
10 0.2 Polymethylpentene
$61.59CAD / Each
Thermo Scientific Nalgene 3663-0025 Clear PMP Graduated Cylinder, 25 mL, 1/Pk 0613420 Mfr # 3663-0025
COMPARE
25 0.5 Polymethylpentene
$68.51CAD / Each
Thermo Scientific Nalgene 3663-0050 Clear PMP Graduated Cylinder, 50 mL, 1/Pk 0613430 Mfr # 3663-0050
COMPARE
50 1 Polymethylpentene
$79.27CAD / Each
Thermo Scientific Nalgene 3663-0100 Clear PMP Graduated Cylinder, 100 mL, 1/Pk 0613440 Mfr # 3663-0100
COMPARE
100 1 Polymethylpentene
$79.27CAD / Each
Thermo Scientific Nalgene 3663-0250 Clear PMP Graduated Cylinder, 250 mL, 1/Pk 0613450 Mfr # 3663-0250
COMPARE
250 2 Polymethylpentene
$110.21CAD / Each
Thermo Scientific Nalgene 3663-0500 Clear PMP Graduated Cylinder, 500 mL, 1/Pk 0613460 Mfr # 3663-0500
COMPARE
500 5 Polymethylpentene
$132.31CAD / Pkg of 1
Thermo Scientific Nalgene 3663-1000 Clear PMP Graduated Cylinder, 1000 mL 0613470 Mfr # 3663-1000
COMPARE
1000 10 Polymethylpentene
$185.65CAD / Each
Thermo Scientific Nalgene 3663-2000 Clear PMP Graduated Cylinder, 2000 mL, 1/Pk 0613480 Mfr # 3663-2000
COMPARE
2000 20 Polymethylpentene
$277.00CAD / Each
Thermo Scientific Nalgene 3663-4000 Clear PMP Graduated Cylinder, 4000 mL, 1/Pk 0613490 Mfr # 3663-4000
COMPARE
4000 50 Polymethylpentene
$626.19CAD / Each
Showing 1- 9 of 9