Thermo Scientific Nalgene Calibrated Graduated Cylinders, PP
Polypropylene (PP) cylinders offer very good chemical resistance
  • Resists temperatures up to 135°C (275°F) for a variety of uses
  • Translucent cylinder with blue graduations for easy visibility
  • Meet ASTM Class B, E1272 specifications and ISO Standard 6706
Polypropylene (PP) cylinders have molded graduations. Calibrated T.C./T.D. at 20°C. Nonwetting; no meniscus. Large, stable base. Autoclavable, but may affect accuracy. Call for additional sizes.
$56.43 - $722.70CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Thermo Scientific Nalgene 3662-0010 Polypropylene Graduated Cylinder, 10 mL, 1/Pk 0613510 Mfr # 3662-0010
COMPARE
10 0.2 Polypropylene
$56.43CAD / Each
Thermo Scientific Nalgene 3662-0025 Polypropylene Graduated Cylinder, 25 mL, 1/Pk 0613520 Mfr # 3662-0025
COMPARE
25 0.5 Polypropylene
$63.06CAD / Each
Thermo Scientific Nalgene 3662-0050 Polypropylene Graduated Cylinder, 50 mL 0613530 Mfr # 3662-0050
COMPARE
50 1 Polypropylene
$71.16CAD / Each
Thermo Scientific Nalgene 3662-0100 Polypropylene Graduated Cylinder, 100 mL, 1/Pk 0613540 Mfr # 3662-0100
COMPARE
100 1 Polypropylene
$79.27CAD / Each
Thermo Scientific Nalgene 3662-0250 Polypropylene Graduated Cylinder, 250 mL, 1/Pk 0613550 Mfr # 3662-0250
COMPARE
250 2 Polypropylene
$98.72CAD / Each
Thermo Scientific Nalgene 3662-0500 Polypropylene Graduated Cylinder, 500 mL, 1/Pk 0613560 Mfr # 3662-0500
COMPARE
500 5 Polypropylene
$137.03CAD / Pkg of 1
Thermo Scientific Nalgene 3662-1000 Polypropylene Graduated Cylinder, 1000 mL, 1/Pk 0613570 Mfr # 3662-1000
COMPARE
1000 10 Polypropylene
$204.43CAD / Each
Thermo Scientific Nalgene 3662-2000 Polypropylene Graduated Cylinder, 2000 mL, 1/Pk 0613580 Mfr # 3662-2000
COMPARE
2000 20 Polypropylene
$277.00CAD / Each
Thermo Scientific Nalgene 3662-4000 Polypropylene Graduated Cylinder, 4000 mL, 1/Pk 0613590 Mfr # 3662-4000
COMPARE
4000 50 Polypropylene
$722.70CAD / Each
Showing 1- 9 of 9