Thermo Scientific Lindberg Blue M Mini-Mite Tube Furnaces
Compact, portable, and lightweight design make furnaces ideal for your benchtop
  • Excellent temperature control prevents set point overshoot
These small, reliable tube furnaces meet the specific needs of educational, governmental, and industrial laboratories. Choose from furnaces with single set point or 16-segment programmable controllers.
All models feature microprocessor-based, self-tuning PID control to prevent overshooting of set temperature. As a safety precaution, power to the heating elements is cut off when the chamber temperature exceeds the set temperature or when the door is opened. Overtemperature protection is adjustable—set according to your application. The LED lets you view set temperature and actual temperature simultaneously.
Split hinge design permits convenient placement of the process tube (process tube not included—order separately). Chamber is insulated with energy-efficient Moldatherm® for quick heat up and cool down rates.
$5,613.62 - $6,365.04CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Control Type Power (VAC) Process Tube Length (in)
Availability Pricing
Thermo Scientific Lindberg TF55035A-1 Blue M Mini-Mite Tube Furnace, 16-Pt; 120V 3385000 Mfr # TF55035A-1
COMPARE
16-segment 120 12
$6,365.04CAD / Each
Thermo Scientific Lindberg TF55030C-1 Blue M Mini-Mite Tube Furnace, SSP; 208V 3385015 Mfr # TF55030C-1
COMPARE
Single set point 208 / 240 12
$5,613.62CAD / Each