Thermo Scientific™ Hypersil™ ODS Classical C18 HPLC Columns
High-efficiency columns with proven results and reproducibility
  • Batch tested to guarantee your results
  • Separate a wide range of compounds
Thermo Scientific™ Hypersil™ columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil ODS C18 Columns are exceptionally reliable and capable of separating a wide range of compounds including nonpolar, moderately polar, and lipophilic compounds such as triglycerides. Stable in pH range of 2 to 8.
$1,407.09 - $2,799.44CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific 30103-054630 ODS HPLC Column, C18, 3 µm, 50 mm L x 4.6 mm ID 0188724 Mfr # 30103-054630
COMPARE
4.6 50 3
$1,427.71CAD / Each
Thermo Scientific 30103-104030 ODS (C18) Column - Length 100mm, 4.0mm ID, 3µm particle size 0188727 Mfr # 30103-104030
COMPARE
4 100 3
$1,539.69CAD / Each
Thermo Scientific Hypersil 30103-104630 ODS HPLC Column, C18, 3 µm, 100 mm L x 4.6 mm ID 0188728 Mfr # 30103-104630
COMPARE
4.6 100 3
$1,532.33CAD / Each
Thermo Scientific 30103-154630 ODS HPLC Column, C18, 3 µm, 150 mm L x 4.6 mm ID 0188736 Mfr # 30103-154630
COMPARE
4.6 150 3
$2,799.44CAD / Each
Thermo Scientific 30103-254630 ODS HPLC Column, C18, 3 µm, 250 mm L x 4.6 mm ID 0188740 Mfr # 30103-254630
COMPARE
4.6 250 3
$1,654.62CAD / Each
Thermo Scientific 30105-054630 ODS HPLC Column, C18, 5 µm, 50 mm L x 4.6 mm ID 0188748 Mfr # 30105-054630
COMPARE
4.6 50 5
$1,488.12CAD / Each
Thermo Scientific 30105-102130 ODS HPLC Column, C18, 5 µm, 100 mm L x 2.1 mm ID 0188749 Mfr # 30105-102130
COMPARE
2.1 100 5
$1,701.77CAD / Each
Thermo Scientific 30105-104630 ODS HPLC Column, C18, 5 µm, 100 mm L x 4.6 mm ID 0188752 Mfr # 30105-104630
COMPARE
4.6 100 5
$1,407.09CAD / Each
Thermo Scientific 30105-124030 ODS HPLC Column, C18, 5 µm, 125 mm L x 4.0 mm ID 0188755 Mfr # 30105-124030
COMPARE
4 125 5
$1,944.87CAD / Each
Thermo Scientific 30105-124630 ODS HPLC Column, C18, 5 µm, 125 mm L x 4.6 mm ID 0188756 Mfr # 30105-124630
COMPARE
4.6 125 5
$2,453.19CAD / Each
Thermo Scientific 30105-154630 ODS HPLC Column, C18, 5 µm, 150 mm L x 4.6 mm ID 0188760 Mfr # 30105-154630
COMPARE
4.6 150 5
$2,342.69CAD / Each
Thermo Scientific 30105-204630 ODS (C18) Column - Length 200mm, 4.6mm ID, 5µm particle size 0188764 Mfr # 30105-204630
COMPARE
4.6 200 5
$1,539.69CAD / Each
Thermo Scientific 30105-252130 ODS (C18) Column - Length 250mm, 2.1mm ID, 5µm particle size 0188765 Mfr # 30105-252130
COMPARE
2.1 250 5
$1,620.73CAD / Each
Thermo Scientific Hypersil 30105-254630 ODS HPLC Column, C18, 5 µm, 250 mm L x 4.6 mm ID 0188768 Mfr # 30105-254630
COMPARE
4.6 250 5
$1,498.44CAD / Each
Thermo Scientific Hypersil 30110-254630 ODS (C18) Column - Length 250mm, 4.6mm ID, 10µm particle size 0188769 Mfr # 30110-254630
COMPARE
4.6 250 10
$1,410.03CAD / Each
Showing 1- 15 of 15