Thermo Scientific Cimarec Multiposition Electromagnetic Stirrers
No moving parts for quiet, maintenance-free operation
  • Adjustable wattage is ideal for mixing thermally unstable media
Select either a nonimmersible stirrer with adjustable wattage or an immersible stirrer with fixed or adjustable wattage. All are available with 6 or 15 stirring positions to simultaneously stir at precisely the same speed. 60 position stirrers are immersible. Touch the stir key, and the stir bars automatically center and start to stir. Stirrers feature user interface in English, French, German, and Spanish.
All stirrers are hermetically sealed in a stainless steel housing to protect against moisture, spills, corrosion, dust and dirt. All stirrers are supplied with a 6-ft cord with plug.
Non-Immersible Stirrers let you set the wattage at 5, 10, 15, or 20 watts. Use a lower setting for long term operation; use a higher setting for large volumes and viscous solutions. Set speed and wattage by simply pressing the positive or negative key on the keypad. A bright 4-digit LED shows speed and wattage setting. These stirrers feature IP68 rating and are hermetically sealed to protect against moisture, spills, corrosion, dust and dirt.
Immersible Stirrers are available with 7-, 20-, or 40-watt power level. Adjust the power usage on the 20-watt models at 5, 10, 15 or 20 watts; adjust the power usage on the 40-watt models at 4, 8, 12, 16, 20, 24, 28, 32, 36, or 40 watts. The immersible stirrers fit directly in the clear PLEXIGLAS® tank 01137-20 sold below; tank seams are welded to prevent leaking.
Program the 20-watt immersible stirrers to continuously stir or to alternately stir and rest at selected time intervals. Stirring time is adjustable from 8 seconds to 8 minutes; resting time is adjustable from 4 seconds to 4 minutes. Control 40-watt stirrers via your computer with the built-in RS-232 interface.
$4,331.77 - $9,709.64CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
No. Of Stirring Positions Power (VAC)
Availability Pricing
Thermo Scientific Cimarec 50088077 iTelesystem 6-Position Stirrer w20C Control 0465015 Mfr # 50088077
COMPARE
6 100 to 240
$5,083.20CAD / Each
Thermo Scientific Cimarec 50088034 iTelesystem 15-Position Stirrer w20C Control 0465025 Mfr # 50088034
COMPARE
15 100 to 240
$6,556.59CAD / Each
Thermo Scientific Cimarec 50093557 Multipoint 6 Non-Immersible Stirrer 0465110 Mfr # 50093557
COMPARE
6 100 to 240
$4,331.77CAD / Each
Thermo Scientific Cimarec 50093538 Multipoint 15 Non-Immersible Stirrer; US Plug 0465120 Mfr # 50093538
COMPARE
15 100 to 240
$6,026.17CAD / Each
Thermo Scientific Cimarec 50088036 iTelesystem 15-Position Stirrer w40C Control 0465140 Mfr # 50088036
COMPARE
15 115
$7,985.77CAD / Each
Thermo Scientific Cimarec 50088078 iTelesystem 6-Position Stirrer w40C Control 0465161 Mfr # 50088078
COMPARE
6 100 to 240
$6,365.04CAD / Each
Thermo Scientific Cimarec 50088009 iTelesystem 60-Position Stirrer w20C Control 0465167 Mfr # 50088009
COMPARE
60 115 / 230
$9,709.64CAD / Each
Showing 1- 7 of 7