Thermo Scientific Cimarec+ Hot Plates
High precision temperature control and exceptional safety
  • Adjust temperature in one degree increments on the easy-to-read LED display
  • Hot top indicator alerts when the plate temperature is above 50°C
  • Raised display design prevents damage to electronics from spills
  • Automatic overtemperature protection cuts power to allow cool down
  • Choose aluminum for more even temperature distribution or ceramic for higher temperatures and chemical resistance
The Cimarec+ series of hot plates combines user friendly analog temperature control with a more accurate digital display. The hot top indicator remains lit as long as the plate temperature is above 50°C, even if the hot plate is turned off. Optional external temperature probe (order separately) provides the feedback necessary to keep your sample at the desired temperature. Optional splash guard (order separately) protects you and your sample from mishaps. Optional rod and clamps (order separately) provide the means to secure your sample container to the top plate.
$630.61 - $1,301.00CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Length (in) Top Plate Material
Availability Pricing
Thermo Scientific Cimarec HP88854100 Hot Plate, 4x4" Ceramic; 120 VAC 0460001 Mfr # HP88854100
COMPARE
120 4 Ceramic
$701.33CAD / Each
Thermo Scientific Cimarec HP88854105 Hot Plate, 4x4" Ceramic; 230 VAC 0460002 Mfr # HP88854105
COMPARE
230 4 Ceramic
$630.61CAD / Each
Thermo Scientific Cimarec HP88857100 Hot Plate, 7x7" Ceramic; 120 VAC 0460007 Mfr # HP88857100
COMPARE
120 7 Ceramic
$686.60CAD / Each
Thermo Scientific Cimarec HP88857105 Hot Plate, 7x7" Ceramic; 230 VAC 0460008 Mfr # HP88857105
COMPARE
230 7 Ceramic
$761.74CAD / Each
Thermo Scientific Cimarec HP88857104 Hot Plate, 7x7" Aluminum; 120 VAC 0460016 Mfr # HP88857104
COMPARE
120 7 Aluminum
$886.98CAD / Each
Thermo Scientific Cimarec HP88850100 Hot Plate, 10x10" Ceramic; 120 VAC 0460040 Mfr # HP88850100
COMPARE
120 10 Ceramic
$1,138.93CAD / Each
Thermo Scientific Cimarec HP88850105 Hot Plate, 10x10" Ceramic; 230 VAC 0460041 Mfr # HP88850105
COMPARE
230 10 Ceramic
$1,301.00CAD / Each
Showing 1- 7 of 7