Thermo Scientific Barnstead Cartridge Kits for use with E-pure Systems
Select pretreatment, deionization, or organic-free cartridges, depending upon
your application
  • For use with E-Pure systems
  • Produce MΩ-cm resistivity, Type I reagentgrade water
Cartridge kits include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagentgrade water. Kits that include organic-free cartridges produce water that has less than 10 ppb
organic carbon. Kits for Nanopure Infinity and Nanopure Dlamond systems include final filter; order final filter for quad-style Nanopure and E-Pure systems separately.
$1,029.90 - $1,561.79CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific Barnstead D5029 Type I cartridge kit, three module RK-99258-11 Mfr # D5029
COMPARE
Type I cartridge kit, three module
$1,029.90CAD / Each
Thermo Scientific Barnstead D5022 Type I cartridge kit, organic free, three module RK-99258-12 Mfr # D5022
COMPARE
Type I cartridge kit, organic free, three module
$1,561.79CAD / Each
Thermo Scientific Barnstead D5028 Type I cartridge kit, four module RK-99258-21 Mfr # D5028
COMPARE
Type I cartridge kit, four module
$1,539.69CAD / Each
Thermo Scientific Barnstead D5023 Type I cartridge kit, organic free, four module RK-99258-22 Mfr # D5023
COMPARE
Type I cartridge kit, organic free, four module
$1,554.43CAD / Each