Thermo Scientific AquaSensors DataStick Turbidity Sensor
Plug-and-play MODBUS RTU communications for direct output of measured readings
  • Wide 0 to 4000 NTU measurement range — ideal for drinking water applications!
  • Infrared light source conforms to ISO 7027 standard
Thermo Scientific AquaSensors DataStick features rugged construction for on-line turbidity measurement in process environments. Durable polycarbonate sensor head has exceptional chemical resistance and mechanical strength. CPVC back body and communications adapter features 1" NPT(M) threads for submersible installation. Sensor comes precalibrated for turbidity and temperature for easy initial setup and reading.
DataStick features MODBUS plug-and-play communications for direct data reporting. Optional local display provides easy viewing of measured readings. Display comes with a 4 to 20 mA output for relay of measurements to a PLC, datalogger, or chart recorder. Other output configurations are available via special order.
$6,632.24CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Control Type Output Current
Availability Pricing
Thermo Scientific DataStick Wide-Range Turbidity Sensor 9990010 Mfr # RT1141
COMPARE
On/Off
$6,632.24CAD / Each