Test Tube Brushes
These brushes are specificaly designed for cleaning test tubes. Radial Tip brushes have formed brush sprays at the tip allowing thorough cleaning of all sides. Stainless steel wire stem Radial Tip brushes are extremely durable and provide superior performance. Tied Tip brushes are designed to enter small openings of bottles and tubes, then expand to clean the sides and bottom without scratching . Sponge Tip are made to expand to give a more thorough cleaning while also protecting from scratches. The adjustable test tube brush spreads apart to fit all sizes of test tubes.
$25.78 - $137.17CAD / Pkg of 12
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (in) Handle Length (in) Material
Availability Pricing
COMPARE
0.5 8 Nylon
$108.59CAD / Pkg of 12
COMPARE
1 10 Nylon
$96.51CAD / Pkg of 12
COMPARE
1.625 9 Nylon
$107.41CAD / Pkg of 12
COMPARE
0.625 10 Nylon
$75.29CAD / Pkg of 12
COMPARE
0.875 10 Natural bristles, wire handle
$77.79CAD / Pkg of 12
COMPARE
1.375 11 Natural bristles, wire handle
$104.61CAD / Pkg of 12
COMPARE
0.5 8 Nylon
$82.95CAD / Pkg of 12
COMPARE
0.75 9 Nylon
$97.24CAD / Pkg of 12
COMPARE
1.375 10 Nylon
$108.29CAD / Pkg of 12
COMPARE
1.375 9 Nylon
$108.29CAD / Pkg of 12
COMPARE
1.5 10 Nylon
$109.33CAD / Pkg of 12
COMPARE
0.5 7 Natural bristles, wire handle
$137.17CAD / Pkg of 12
COMPARE
0.75 6 Natural bristles, wire handle
$123.03CAD / Pkg of 12
COMPARE
1.25 9 Natural bristles, wire handle
$124.21CAD / Pkg of 12
COMPARE
1.5 1 Nylon
$25.78CAD / Each
Showing 1- 15 of 15