Taylor Wireless Digital Food Service Thermometers
Monitor food cooking temperatures without opening the oven door
  • Use the magnet on the back of the base to mount to a stove while the remote probe monitors temperatures in the oven
  • Combination thermometer/timer with pre-programmed meat cooking temperatures
  • Thermometer alarms when timer reaches zero or when food is done cooking
The real secret to great cooking is a Taylor thermometer. Thermometers are the only way to ensure that foods are cooked to the proper temperature, your oven is calibrated to the correct degree, and that your refrigerator is keeping your food cold and safe. Taylor is and has been the leader in food thermometers, with expertise over 150 years strong. You can count on Taylor for accuracy, superior performance, and dependability. That is why Taylor is tools chef's trust. This digital thermometer with wireless remote for monitoring cooking temperatures for up to 200' away. The base unit has stand or magnet positioning. Remote unit has lanyard, clip, stand or magnet positioning.
$59.23CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Max Temperature (° F) Description
Availability Pricing
Taylor 1479-21 Pro Series Wireless Cooking Thermometer with Timer and Pager 9000116 Mfr # 147921
COMPARE
232 450 Wireless Cooking Thermometer with Timer and Pager
$59.23CAD / Each