Sub-Micro Quartz Cuvettes for Absorbance Spectroscopy
Fits most standard-sized (10 mm) cuvette sample holders
  • Constructed of durable quartz
Available for UV/Vis absorbance spectroscopy applications (two polished windows). Match the cuvette center height to your instrument's light beam height; see individual cuvette specifications to verify correct manufacturer beam height.
$1,797.54 - $2,254.29CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Beam Height Zd (mm) Max Capacity (MicroL) Pathlength (mm)
Availability Pricing
COMPARE
8.5 10 10
$1,915.41CAD / Each
COMPARE
15 10 10
$1,915.41CAD / Each
COMPARE
8.5 50 10
$2,254.29CAD / Each
COMPARE
8.5 100 10
$1,797.54CAD / Each
COMPARE
15 100 10
$1,797.54CAD / Each