Speirs Robertson AMCU Series High-Capacity Instrument Isolation Platforms
Improves performance of sensitive instruments by shielding them from vibrations
  • Isolates heavy instruments from vibrations
  • High performance 3Hz natural frequency cushion air isolators
  • Suitable for instruments from 200kg up to 1000kgs
Save time and money by making sure your expensive high performance instruments are not underperforming due to a poor working environment. These platforms feature four internal air cushion isolators which are pumped up using a foot pump until the platform top floats free from the floor, so isolating the top from vibrations below. The stainless worktop is strong and durable and can be easily cleaned. The low profile of these platforms allows them to be installed easily into most areas. Platforms are simple to install, easy to use and maintenance free.
$7,091.43 - $8,060.92CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Height (in) Width (in) Description
Availability Pricing
Speirs Robertson AMCU-75x75 AMCU Instrument Platform, Sorbothane Isolators; 75x75 cm 1150023 Mfr # AMCU-75X75
COMPARE
4 30 AMCU Instrument Platform, Sorbothane Isolators; 75x75 cm
$7,529.02CAD / Each
Speirs Robertson AMCU-90x75 AMCU Instrument Platform, Sorbothane Isolators; 90x75 cm 1150024 Mfr # AMCU-90X75
COMPARE
4 36 AMCU Instrument Platform, Sorbothane Isolators; 90x75 cm
$7,091.43CAD / Each
Speirs Robertson AMCU-90x90 AMCU Instrument Platform, Sorbothane Isolators; 90x90 cm 1150025 Mfr # AMCU-90X90
COMPARE
4 36 AMCU Instrument Platform, Sorbothane Isolators; 90x90 cm
$8,060.92CAD / Each