SHOWA N-DEX Plus Industrial Grade Nitrile Gloves
Reduce hand fatigue by keeping your hands cool and dry
  • Nonlatex—made of soft 100% nitrile
  • Three times the puncture-resistance of natural rubber or vinyl
  • Superior tactile-sensitivity, comfort, and dexterity
  • Lightweight and disposable
N-DEX® gloves are made from a special low-modulus, nonlatex, soft nitrile formulation. They offer greater tactile-sensitivity, comfort, dexterity, and puncture-resistance than latex gloves. All N-DEX® nitrile gloves are ambidextrous and blue in color.
Industrial Grade N-DEX® Plus Gloves are thicker for greater puncture and tear resistance. Gloves provide good resistance to chemicals and abrasion. Ideal for tough lab conditions, assembly, or manufacturing. Available only in low-powder versions. Box of 50.
$36.83 - $47.89CAD / Pkg of 50
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
SHOWA N-DEX Industrial Grade Nitrile Gloves, Size S; 50/PK RK-86287-60 Mfr # 8005S
COMPARE
9 1/2 Small
$47.89CAD / Pkg of 50
SHOWA N-DEX Industrial Grade Nitrile Gloves, Size M; 50/PK RK-86287-61 Mfr # 8005M
COMPARE
9 1/2 Medium
$45.82CAD / Pkg of 50
SHOWA N-DEX Industrial Grade Nitrile Gloves, Size L; 50/PK RK-86287-62 Mfr # 8005L
COMPARE
9 1/2 Large
$41.11CAD / Pkg of 50
SHOWA N-DEX Industrial Grade Nitrile Gloves, Size XL; 50/PK RK-86287-63 Mfr # 8005XL
COMPARE
9 1/2 X-Large
$36.83CAD / Pkg of 50