Shimpo TTC Digital Torque Tool Testers
Quick and accurate testing
  • Analog bar graph and pass/fail icons provide instant results
  • Fast sampling rate ensures an accurate test
  • Standard USB and RS-232 outputs allow download of data to PC
These digital torque tool testers feature enhanced versatility to fit into any testing environment. The light-weight, compact aluminum housing is durable and portable. The tester runs on rechargeable batteries or AC power (both included), so you can plug into the wall to save battery or bring it with you. Backlit, reversible, graphical display with analog-style bar graph and pass/fail icons produce immediate results in any lighting conditions.
Expansive 1000-point memory stores test data for later download to PC via the USB or RS-232 output. Six modes of operation offer different test parameters: Track for real-time readings, Peak records the highest reading over the course of a test, First Peak records the highest reading after a programmed decrease, Auto Peak records highest reading after a period of time, Auto First Peak resets the first peak after a period of time, and Double Peak records two separate peak readings from programmed decreases.
$1,510.22 - $3,388.80CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-in) Capacity (N-m)
Availability Pricing
Shimpo TTC-I-2 Digital Torque Tester with Internal Sensor, 2 N-m Capacity 2530424 Mfr # TTC-I-2
COMPARE
17.7 2
$1,510.22CAD / Each
Shimpo TTC-I-5 Digital Torque Tester with Internal Sensor, 5 N-m Capacity 2530425 Mfr # TTC-I-5
COMPARE
43.3 5
$1,510.22CAD / Each
Shimpo TTC-I-10 Digital Torque Tester with Internal Sensor, 10 N-m Capacity 2530426 Mfr # TTC-I-10
COMPARE
88.5 10
$2,151.15CAD / Each
Shimpo TTC-I-20 Digital Torque Tester with Internal Sensor, 20 N-m Capacity 2530427 Mfr # TTC-I-20
COMPARE
177 20
$1,804.90CAD / Each
Shimpo TTC-I-50 Digital Torque Tester with Internal Sensor, 50 N-m Capacity 2530428 Mfr # TTC-I-50
COMPARE
443 50
$1,989.08CAD / Each
Shimpo TTC-I-100  Digital Torque Tester with Internal Sensor, 100 N-m Capacity 2530429 Mfr # TTC-I-100
COMPARE
885 100
$2,246.92CAD / Each
Shimpo TTC-E-200 Digital Torque Tester with External Sensor, 200 N-m 2530430 Mfr # TTC-E-200
COMPARE
1770 200
$3,094.12CAD / Each
Shimpo TTC-E-500 Digital Torque Tester with External Sensor, 500 N-m 2530431 Mfr # TTC-E-500
COMPARE
4425 500
$3,388.80CAD / Each
Showing 1- 8 of 8