Scienceware Sampling Spoons, PS

Reduce cross-contamination
  • Ideal for single use
  • Made of high-impact polystyrene
High-impact polystyrene sampling spoons are precise enough to collect specific volume amounts and economical enough to use a separate spoon for each substance — reducing your chances of cross-contamination.
$36.39 - $44.64CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Sterile Capacity (mL)
Availability Pricing
Scienceware F36721-0005 Polystyrene Sampling Spoons, 0.05 mL Capacity; 25/Pk 0626350 Mfr # F36721-0005
COMPARE
Polystyrene No 0.05
$44.64CAD / Each
Scienceware F36721-0010 Polystyrene Sampling Spoons, 0.10 mL Capacity; 25/Pk 0626352 Mfr # F36721-0010
COMPARE
Polystyrene No 0.1
$36.39CAD / Each
Scienceware F36721-0025 Polystyrene Sampling Spoons, 0.25 mL Capacity; 25/Pk 0626354 Mfr # F36721-0025
COMPARE
Polystyrene No 0.25
$37.13CAD / Each
Scienceware F36721-0050 Polystyrene Sampling Spoons, 0.50 mL Capacity; 25/Pk 0626356 Mfr # F36721-0050
COMPARE
Polystyrene No 0.5
$37.13CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat