Scienceware Lab-Aire II Drying Racks
Available in single or double-sided, benchtop or wall-mount, with up to 114 peg positions
Use these high-capacity racks to safely dry and store frequently used or fragile labware. Rugged, white ABS plastic body and contrasting blue pegs will never corrode and are easy to clean. Racks come with three sizes of V-channel pegs that can be repositioned to hold a variety of labware. Drip tray has molded-in scalloped wells to accommodate long, thin labware such as burettes and pipettes.
Single-sided models can be placed against a wall, conserving bench space. Double-sided freestanding models provide twice the capacity with only a slightly larger footprint. Wall-mount models have mounting points sized to fit standard pegboards; hardware not included.
$317.15 - $671.87CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Tubes Held Description
Availability Pricing
Scienceware f18933-0012 Drying Rack, Benchtop, single-sided 2 tier 0673393 Mfr # F18933-0012
COMPARE
16 1/4 38 Drying Rack, Benchtop, single-sided 2 tier
$345.14CAD / Each
Scienceware F18933-0015 Drying Rack, Benchtop, single-sided 3 tier 0673394 Mfr # F18933-0015
COMPARE
23 3/4 57 Drying Rack, Benchtop, single-sided 3 tier
$435.02CAD / Each
Scienceware F18933-0021 Drying Rack, Benchtop, double-sided 1 tier 0673395 Mfr # F18933-0021
COMPARE
16 1/4 38 Drying Rack, Benchtop, double-sided 1 tier
$317.15CAD / Each
Scienceware F18933-0022 Drying Rack, Benchtop, double-sided 2 tier 0673396 Mfr # F18933-0022
COMPARE
16 1/4 76 Drying Rack, Benchtop, double-sided 2 tier
$487.69CAD / Each
Scienceware F18933-0023 Drying Rack, Benchtop, double-sided 3 tier 0673397 Mfr # F18933-0023
COMPARE
23 3/4 114 Drying Rack, Benchtop, double-sided 3 tier
$671.87CAD / Each
Scienceware F18933-0013 Drying Rack, Wall-mount, single-sided 3 tier 0673399 Mfr # F18933-0013
COMPARE
23 3/4 57 Drying Rack, Wall-mount, single-sided 3 tier
$328.20CAD / Each