Scienceware Graduated Cylinders with Handle, PP
The handle on these polypropylene cylinders provides a solid grip, making pouring safe and easy. The hand opening on the handle measures 3-1/2'' x 1-1/8''. Cylinders have a double scale with raised graduations for easy reading. Sturdy hexagonal base. Steam autoclavable at 250degF (121degC). Bel Art.
$49.21 - $134.37CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Scienceware 28461-1000 graduated polypropylene cylinder with handle, 1000 mL 0613710 Mfr # 28461-1000
COMPARE
1000 10 Polypropylene
$49.21CAD / Each
Scienceware 28461-2000 graduated polypropylene cylinder with handle, 2000 mL 0613720 Mfr # 28461-2000
COMPARE
2000 20 Polypropylene
$134.37CAD / Each